Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 30920452-30920725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAN1
Entrez Gene ID
22909
Gene Name
FANCD2/FANCI-associated nuclease 1
Gene Aliases
KIAA1018, KMIN, MTMR15, hFAN1
UniGene
Hs.584863
Gene Chromosome Location
Chr.15: 30903695-30943108 on Build GRCh38
Transcripts
NM_014967.4 XM_005254232.4 XM_005254234.4 XM_005254235.3 XM_011521370.2 XM_011521371.1 XM_017022012.1 XM_017022013.1 XM_011521737.2

Gene Symbol
MTMR10
Entrez Gene ID
54893
Gene Name
myotubularin related protein 10
Gene Aliases
-
UniGene
Hs.30141
Gene Chromosome Location
Chr.15: 30919764-30991604 on Build GRCh38
Transcripts
NM_014967.4 XM_005254232.4 XM_005254234.4 XM_005254235.3 XM_011521370.2 XM_011521371.1 XM_017022012.1 XM_017022013.1 XM_011521737.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
960358 c.2002A>G p.R668G Chr.15: 30920603-30920603 on Build GRCh38
236640 c.2041C>A p.H681N Chr.15: 30920642-30920642 on Build GRCh38
3420224 c.2008T>G p.L670V Chr.15: 30920609-30920609 on Build GRCh38
6660399 c.2015G>A p.R672Q Chr.15: 30920616-30920616 on Build GRCh38
4391145 c.2034G>C p.Q678H Chr.15: 30920635-30920635 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758707862 Chr.15: 30920524-30920524 on Build GRCh38
rs755918280 Chr.15: 30920630-30920630 on Build GRCh38
rs761818803 Chr.15: 30920615-30920615 on Build GRCh38
rs372222101 Chr.15: 30920501-30920501 on Build GRCh38
rs560877684 Chr.15: 30920660-30920660 on Build GRCh38
rs768342788 Chr.15: 30920696-30920696 on Build GRCh38
rs920053576 Chr.15: 30920478-30920478 on Build GRCh38
rs761906714 Chr.15: 30920496-30920496 on Build GRCh38
rs949660925 Chr.15: 30920706-30920706 on Build GRCh38
rs376651205 Chr.15: 30920686-30920686 on Build GRCh38
rs562747667 Chr.15: 30920684-30920684 on Build GRCh38
rs200818425 Chr.15: 30920574-30920574 on Build GRCh38
rs745572671 Chr.15: 30920552-30920552 on Build GRCh38
rs750653191 Chr.15: 30920514-30920514 on Build GRCh38
rs76554225 Chr.15: 30920698-30920698 on Build GRCh38
rs368477312 Chr.15: 30920502-30920506 on Build GRCh38
rs766624148 Chr.15: 30920661-30920661 on Build GRCh38
rs753269805 Chr.15: 30920673-30920673 on Build GRCh38
rs186701091 Chr.15: 30920669-30920669 on Build GRCh38
rs369345963 Chr.15: 30920668-30920668 on Build GRCh38
rs748145720 Chr.15: 30920699-30920699 on Build GRCh38
rs765185424 Chr.15: 30920499-30920503 on Build GRCh38
rs369028560 Chr.15: 30920499-30920499 on Build GRCh38
rs376833677 Chr.15: 30920531-30920531 on Build GRCh38
rs200971774 Chr.15: 30920541-30920541 on Build GRCh38
rs748791115 Chr.15: 30920548-30920548 on Build GRCh38
rs149953272 Chr.15: 30920549-30920549 on Build GRCh38
rs767355576 Chr.15: 30920706-30920707 on Build GRCh38
rs940671449 Chr.15: 30920596-30920596 on Build GRCh38
rs10163148 Chr.15: 30920481-30920481 on Build GRCh38
rs751551936 Chr.15: 30920586-30920595 on Build GRCh38
rs766825197 Chr.15: 30920530-30920530 on Build GRCh38
rs952154809 Chr.15: 30920571-30920571 on Build GRCh38
rs765317735 Chr.15: 30920616-30920616 on Build GRCh38
rs759383731 Chr.15: 30920672-30920673 on Build GRCh38
rs752006055 Chr.15: 30920663-30920663 on Build GRCh38
rs140501687 Chr.15: 30920573-30920573 on Build GRCh38
rs542981453 Chr.15: 30920678-30920678 on Build GRCh38
rs976068372 Chr.15: 30920656-30920656 on Build GRCh38
rs143187330 Chr.15: 30920682-30920682 on Build GRCh38
rs937119347 Chr.15: 30920680-30920680 on Build GRCh38
rs373504503 Chr.15: 30920569-30920569 on Build GRCh38
rs796444081 Chr.15: 30920631-30920631 on Build GRCh38
rs907850473 Chr.15: 30920589-30920589 on Build GRCh38
rs144081053 Chr.15: 30920562-30920562 on Build GRCh38
rs540563961 Chr.15: 30920657-30920657 on Build GRCh38
rs777125944 Chr.15: 30920546-30920546 on Build GRCh38
rs746895584 Chr.15: 30920572-30920572 on Build GRCh38
rs746782057 Chr.15: 30920688-30920688 on Build GRCh38
rs1021956750 Chr.15: 30920513-30920513 on Build GRCh38
rs778650145 Chr.15: 30920551-30920551 on Build GRCh38

View Full Product Details