Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122043839-122044096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL7A
Entrez Gene ID
605
Gene Name
BCL tumor suppressor 7A
Gene Aliases
BCL7
UniGene
Hs.530970
Gene Chromosome Location
Chr.12: 122021886-122062044 on Build GRCh38
Transcripts
NM_001024808.2 NM_020993.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5374432 c.310C>T p.P104S Chr.12: 122043924-122043924 on Build GRCh38
6610766 c.379G>A p.G127S Chr.12: 122043993-122043993 on Build GRCh38
3870959 c.404C>T p.A135V Chr.12: 122044018-122044018 on Build GRCh38
5685172 c.363G>T p.S121S Chr.12: 122043977-122043977 on Build GRCh38
6610764 c.360C>T p.N120N Chr.12: 122043974-122043974 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768969911 Chr.12: 122043909-122043909 on Build GRCh38
rs141114357 Chr.12: 122043959-122043959 on Build GRCh38
rs758586556 Chr.12: 122044040-122044040 on Build GRCh38
rs950340443 Chr.12: 122043990-122043990 on Build GRCh38
rs34821485 Chr.12: 122043973-122043973 on Build GRCh38
rs572767168 Chr.12: 122044030-122044030 on Build GRCh38
rs184713755 Chr.12: 122043917-122043917 on Build GRCh38
rs746847373 Chr.12: 122043869-122043869 on Build GRCh38
rs111620654 Chr.12: 122043992-122043992 on Build GRCh38
rs772389192 Chr.12: 122043916-122043916 on Build GRCh38
rs766450245 Chr.12: 122044007-122044007 on Build GRCh38
rs753902391 Chr.12: 122044009-122044009 on Build GRCh38
rs749400935 Chr.12: 122043967-122043967 on Build GRCh38
rs534509232 Chr.12: 122043883-122043883 on Build GRCh38
rs748394870 Chr.12: 122044036-122044036 on Build GRCh38
rs761150586 Chr.12: 122043988-122043988 on Build GRCh38
rs756158547 Chr.12: 122043960-122043960 on Build GRCh38
rs936296160 Chr.12: 122044062-122044062 on Build GRCh38
rs774529819 Chr.12: 122043910-122043910 on Build GRCh38
rs753534683 Chr.12: 122043969-122043970 on Build GRCh38
rs945199075 Chr.12: 122044016-122044016 on Build GRCh38
rs762546924 Chr.12: 122043946-122043946 on Build GRCh38
rs757387359 Chr.12: 122043985-122043985 on Build GRCh38
rs371454940 Chr.12: 122043879-122043879 on Build GRCh38
rs894215728 Chr.12: 122043880-122043880 on Build GRCh38
rs868424938 Chr.12: 122043958-122043958 on Build GRCh38
rs893600228 Chr.12: 122044002-122044002 on Build GRCh38
rs761441546 Chr.12: 122043997-122043997 on Build GRCh38
rs151108243 Chr.12: 122043956-122043956 on Build GRCh38
rs149849440 Chr.12: 122044023-122044023 on Build GRCh38
rs751249871 Chr.12: 122043953-122043953 on Build GRCh38
rs76573037 Chr.12: 122043998-122043998 on Build GRCh38
rs764719337 Chr.12: 122043926-122043926 on Build GRCh38
rs369731262 Chr.12: 122044057-122044057 on Build GRCh38
rs777412253 Chr.12: 122043865-122043865 on Build GRCh38
rs1020885531 Chr.12: 122043912-122043912 on Build GRCh38
rs144915314 Chr.12: 122043989-122043989 on Build GRCh38
rs747409373 Chr.12: 122044077-122044077 on Build GRCh38
rs574303670 Chr.12: 122043864-122043864 on Build GRCh38
rs144413405 Chr.12: 122043887-122043887 on Build GRCh38
rs538468428 Chr.12: 122043976-122043976 on Build GRCh38
rs763596226 Chr.12: 122043993-122043993 on Build GRCh38
rs201873233 Chr.12: 122044006-122044006 on Build GRCh38
rs528911935 Chr.12: 122043911-122043911 on Build GRCh38
rs781194346 Chr.12: 122043871-122043871 on Build GRCh38
rs375961060 Chr.12: 122043888-122043888 on Build GRCh38
rs768720729 Chr.12: 122043969-122043969 on Build GRCh38
rs761225876 Chr.12: 122043919-122043919 on Build GRCh38
rs200170333 Chr.12: 122043952-122043952 on Build GRCh38
rs752255513 Chr.12: 122043927-122043927 on Build GRCh38
rs1056001334 Chr.12: 122043870-122043870 on Build GRCh38
rs926190828 Chr.12: 122044033-122044033 on Build GRCh38
rs375775431 Chr.12: 122043983-122043983 on Build GRCh38
rs1042276610 Chr.12: 122044017-122044017 on Build GRCh38
rs780258527 Chr.12: 122043966-122043966 on Build GRCh38
rs145823862 Chr.12: 122043999-122043999 on Build GRCh38
rs372943822 Chr.12: 122043977-122043977 on Build GRCh38

View Full Product Details