Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31427851-31428094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX6A2
Entrez Gene ID
1339
Gene Name
cytochrome c oxidase subunit 6A2
Gene Aliases
COX6AH, COXVIAH
UniGene
Hs.250760
Gene Chromosome Location
Chr.16: 31427731-31428428 on Build GRCh38
Transcripts
NM_005205.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6409652 c.199A>C p.I67L Chr.16: 31428026-31428026 on Build GRCh38
5182732 c.197G>A p.R66H Chr.16: 31428028-31428028 on Build GRCh38
5384818 c.178C>T p.R60C Chr.16: 31428047-31428047 on Build GRCh38
4466190 c.141C>T p.S47S Chr.16: 31428084-31428084 on Build GRCh38
7297064 c.153G>A p.S51S Chr.16: 31428072-31428072 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781082127 Chr.16: 31427885-31427885 on Build GRCh38
rs776129970 Chr.16: 31427918-31427919 on Build GRCh38
rs968500689 Chr.16: 31428023-31428023 on Build GRCh38
rs764936247 Chr.16: 31427923-31427925 on Build GRCh38
rs745865460 Chr.16: 31427879-31427888 on Build GRCh38
rs773253151 Chr.16: 31427989-31427990 on Build GRCh38
rs373389146 Chr.16: 31427879-31427879 on Build GRCh38
rs764748820 Chr.16: 31427997-31427997 on Build GRCh38
rs765461993 Chr.16: 31428038-31428038 on Build GRCh38
rs762324971 Chr.16: 31428022-31428022 on Build GRCh38
rs149572007 Chr.16: 31427909-31427910 on Build GRCh38
rs1057446938 Chr.16: 31427949-31427949 on Build GRCh38
rs754857052 Chr.16: 31427880-31427880 on Build GRCh38
rs750662079 Chr.16: 31428070-31428070 on Build GRCh38
rs770483419 Chr.16: 31427902-31427902 on Build GRCh38
rs978580253 Chr.16: 31428028-31428028 on Build GRCh38
rs576532718 Chr.16: 31427995-31427995 on Build GRCh38
rs879171477 Chr.16: 31427920-31427920 on Build GRCh38
rs937404696 Chr.16: 31427939-31427939 on Build GRCh38
rs543950432 Chr.16: 31428071-31428071 on Build GRCh38
rs747749229 Chr.16: 31427932-31427932 on Build GRCh38
rs540244464 Chr.16: 31427989-31427989 on Build GRCh38
rs925338224 Chr.16: 31428029-31428029 on Build GRCh38
rs771976545 Chr.16: 31427908-31427909 on Build GRCh38
rs748837514 Chr.16: 31427894-31427894 on Build GRCh38
rs200013837 Chr.16: 31427911-31427912 on Build GRCh38
rs190969236 Chr.16: 31427992-31427992 on Build GRCh38
rs573032627 Chr.16: 31427983-31427983 on Build GRCh38
rs868798796 Chr.16: 31427924-31427924 on Build GRCh38
rs777248892 Chr.16: 31428008-31428008 on Build GRCh38
rs201073840 Chr.16: 31427891-31427891 on Build GRCh38
rs762702132 Chr.16: 31427936-31427936 on Build GRCh38
rs769575258 Chr.16: 31427893-31427893 on Build GRCh38
rs779931973 Chr.16: 31427870-31427870 on Build GRCh38
rs866114321 Chr.16: 31427960-31427960 on Build GRCh38

View Full Product Details