Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38841134-38841395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf98
Entrez Gene ID
388381
Gene Name
chromosome 17 open reading frame 98
Gene Aliases
-
UniGene
Hs.250182
Gene Chromosome Location
Chr.17: 38835088-38841389 on Build GRCh38
Transcripts
NM_001080465.2

SNP Detail

SNP ID Chromosome Location
rs770799128 Chr.17: 38841249-38841251 on Build GRCh38
rs750554801 Chr.17: 38841183-38841183 on Build GRCh38
rs7210156 Chr.17: 38841208-38841208 on Build GRCh38
rs61752602 Chr.17: 38841250-38841250 on Build GRCh38
rs930921823 Chr.17: 38841174-38841176 on Build GRCh38
rs758257845 Chr.17: 38841203-38841203 on Build GRCh38
rs765683291 Chr.17: 38841164-38841164 on Build GRCh38
rs377402503 Chr.17: 38841330-38841330 on Build GRCh38
rs766541161 Chr.17: 38841268-38841268 on Build GRCh38
rs767826241 Chr.17: 38841180-38841180 on Build GRCh38
rs748063679 Chr.17: 38841197-38841197 on Build GRCh38
rs61750911 Chr.17: 38841189-38841189 on Build GRCh38
rs369977699 Chr.17: 38841312-38841312 on Build GRCh38
rs1018607904 Chr.17: 38841298-38841298 on Build GRCh38
rs753205574 Chr.17: 38841167-38841167 on Build GRCh38
rs373358342 Chr.17: 38841301-38841301 on Build GRCh38
rs866750911 Chr.17: 38841299-38841299 on Build GRCh38
rs748725064 Chr.17: 38841313-38841313 on Build GRCh38
rs1685888 Chr.17: 38841252-38841252 on Build GRCh38
rs777662094 Chr.17: 38841276-38841276 on Build GRCh38
rs767938784 Chr.17: 38841333-38841333 on Build GRCh38
rs763572126 Chr.17: 38841253-38841253 on Build GRCh38
rs750261000 Chr.17: 38841284-38841284 on Build GRCh38
rs1045842305 Chr.17: 38841184-38841184 on Build GRCh38
rs756163697 Chr.17: 38841267-38841267 on Build GRCh38
rs746825476 Chr.17: 38841277-38841277 on Build GRCh38
rs201466719 Chr.17: 38841372-38841372 on Build GRCh38
rs769229195 Chr.17: 38841318-38841318 on Build GRCh38
rs373474691 Chr.17: 38841258-38841258 on Build GRCh38
rs757594639 Chr.17: 38841179-38841179 on Build GRCh38
rs762275873 Chr.17: 38841332-38841332 on Build GRCh38
rs975403556 Chr.17: 38841263-38841263 on Build GRCh38
rs757056522 Chr.17: 38841374-38841374 on Build GRCh38
rs369883104 Chr.17: 38841290-38841290 on Build GRCh38
rs573653158 Chr.17: 38841225-38841225 on Build GRCh38
rs1028367126 Chr.17: 38841293-38841293 on Build GRCh38
rs758405695 Chr.17: 38841274-38841274 on Build GRCh38
rs745411971 Chr.17: 38841241-38841241 on Build GRCh38
rs375513531 Chr.17: 38841260-38841260 on Build GRCh38
rs745463786 Chr.17: 38841305-38841305 on Build GRCh38
rs753994511 Chr.17: 38841339-38841339 on Build GRCh38
rs528140278 Chr.17: 38841323-38841323 on Build GRCh38
rs1685889 Chr.17: 38841251-38841251 on Build GRCh38
rs778722822 Chr.17: 38841190-38841190 on Build GRCh38
rs780824582 Chr.17: 38841303-38841303 on Build GRCh38
rs766483222 Chr.17: 38841336-38841336 on Build GRCh38
rs764461114 Chr.17: 38841176-38841176 on Build GRCh38
rs773702400 Chr.17: 38841335-38841335 on Build GRCh38
rs769317533 Chr.17: 38841306-38841306 on Build GRCh38
rs759022396 Chr.17: 38841161-38841161 on Build GRCh38
rs61750926 Chr.17: 38841228-38841228 on Build GRCh38
rs764046819 Chr.17: 38841365-38841365 on Build GRCh38
rs764504486 Chr.17: 38841257-38841257 on Build GRCh38
rs763230188 Chr.17: 38841171-38841171 on Build GRCh38
rs762871456 Chr.17: 38841211-38841212 on Build GRCh38
rs371239248 Chr.17: 38841239-38841239 on Build GRCh38
rs780795222 Chr.17: 38841342-38841342 on Build GRCh38
rs368681296 Chr.17: 38841177-38841177 on Build GRCh38
rs374779578 Chr.17: 38841244-38841244 on Build GRCh38
rs770615199 Chr.17: 38841231-38841231 on Build GRCh38
rs201194773 Chr.17: 38841259-38841259 on Build GRCh38

View Full Product Details