Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 25236877-25237116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNMT3A
Entrez Gene ID
1788
Gene Name
DNA methyltransferase 3 alpha
Gene Aliases
DNMT3A2, M.HsaIIIA, TBRS
UniGene
Hs.515840
Gene Chromosome Location
Chr.2: 25232961-25342590 on Build GRCh38
Transcripts
NM_001320893.1 NM_022552.4 NM_153759.3 NM_175629.2 NR_135490.1 XM_005264175.4 XM_005264177.4 XM_011532662.2 XM_011532663.2 XM_011532666.2 XM_011532667.2 XM_017003526.1 XM_017003527.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5944897 c.2450A>T p.E817V Chr.2: 25236964-25236964 on Build GRCh38
4385519 c.2422_2437del16 p.T808fs*12 Chr.2: 25236977-25236992 on Build GRCh38
7409533 c.2478+9C>T p.? Chr.2: 25236927-25236927 on Build GRCh38
5944195 c.2418_2422delATCCA p.S807fs*3 Chr.2: 25236992-25236996 on Build GRCh38
231548 c.2409_2478del70 p.R803fs*5 Chr.2: 25236936-25237005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769146992 Chr.2: 25236927-25236927 on Build GRCh38
rs918501371 Chr.2: 25237030-25237030 on Build GRCh38
rs770079872 Chr.2: 25236937-25236937 on Build GRCh38
rs748636848 Chr.2: 25237053-25237053 on Build GRCh38
rs765929513 Chr.2: 25236960-25236960 on Build GRCh38
rs779175988 Chr.2: 25236906-25236906 on Build GRCh38
rs569439863 Chr.2: 25237072-25237072 on Build GRCh38
rs772882388 Chr.2: 25236952-25236952 on Build GRCh38
rs755097282 Chr.2: 25237045-25237045 on Build GRCh38
rs146358501 Chr.2: 25236953-25236953 on Build GRCh38
rs529419548 Chr.2: 25237002-25237002 on Build GRCh38
rs866917013 Chr.2: 25237006-25237006 on Build GRCh38
rs879540431 Chr.2: 25236989-25236989 on Build GRCh38
rs777043930 Chr.2: 25236928-25236928 on Build GRCh38
rs752628411 Chr.2: 25236987-25236987 on Build GRCh38
rs375577092 Chr.2: 25236924-25236924 on Build GRCh38
rs373873045 Chr.2: 25236965-25236965 on Build GRCh38
rs750597155 Chr.2: 25237003-25237003 on Build GRCh38
rs143620117 Chr.2: 25236923-25236923 on Build GRCh38
rs758601570 Chr.2: 25237034-25237034 on Build GRCh38
rs762213449 Chr.2: 25236935-25236935 on Build GRCh38
rs376271785 Chr.2: 25237042-25237042 on Build GRCh38
rs942458220 Chr.2: 25237091-25237091 on Build GRCh38
rs746105605 Chr.2: 25236911-25236911 on Build GRCh38
rs759187608 Chr.2: 25236968-25236968 on Build GRCh38
rs370144254 Chr.2: 25237052-25237052 on Build GRCh38
rs772070354 Chr.2: 25236915-25236915 on Build GRCh38
rs35824014 Chr.2: 25237006-25237006 on Build GRCh38
rs910874835 Chr.2: 25237073-25237073 on Build GRCh38
rs561951805 Chr.2: 25236977-25236977 on Build GRCh38

View Full Product Details