Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 67658591-67658838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYRK2
Entrez Gene ID
8445
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 2
Gene Aliases
-
UniGene
Hs.173135
Gene Chromosome Location
Chr.12: 67648732-67665405 on Build GRCh38
Transcripts
NM_003583.3 NM_006482.2 XM_017020032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3464456 c.1714T>C p.S572P Chr.12: 67658621-67658621 on Build GRCh38
5568592 c.1708T>G p.S570A Chr.12: 67658615-67658615 on Build GRCh38
4679856 c.1727T>C p.L576P Chr.12: 67658634-67658634 on Build GRCh38
4679858 c.1749G>C p.M583I Chr.12: 67658656-67658656 on Build GRCh38
330887 c.1801A>T p.S601C Chr.12: 67658708-67658708 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759557043 Chr.12: 67658750-67658750 on Build GRCh38
rs749972090 Chr.12: 67658649-67658649 on Build GRCh38
rs766231212 Chr.12: 67658668-67658668 on Build GRCh38
rs201919838 Chr.12: 67658650-67658650 on Build GRCh38
rs754855606 Chr.12: 67658686-67658686 on Build GRCh38
rs80279715 Chr.12: 67658704-67658704 on Build GRCh38
rs747908377 Chr.12: 67658691-67658691 on Build GRCh38
rs775950698 Chr.12: 67658726-67658726 on Build GRCh38
rs765971690 Chr.12: 67658747-67658747 on Build GRCh38
rs767318357 Chr.12: 67658752-67658752 on Build GRCh38
rs762690292 Chr.12: 67658744-67658744 on Build GRCh38
rs1002732471 Chr.12: 67658695-67658695 on Build GRCh38
rs755847013 Chr.12: 67658758-67658758 on Build GRCh38
rs542814338 Chr.12: 67658753-67658753 on Build GRCh38
rs759178764 Chr.12: 67658713-67658713 on Build GRCh38
rs777979669 Chr.12: 67658760-67658760 on Build GRCh38
rs769580168 Chr.12: 67658734-67658734 on Build GRCh38
rs746447064 Chr.12: 67658724-67658724 on Build GRCh38
rs772319729 Chr.12: 67658761-67658761 on Build GRCh38
rs764940972 Chr.12: 67658636-67658636 on Build GRCh38
rs368545205 Chr.12: 67658722-67658722 on Build GRCh38
rs143848143 Chr.12: 67658629-67658629 on Build GRCh38
rs781089384 Chr.12: 67658689-67658689 on Build GRCh38
rs557417303 Chr.12: 67658720-67658720 on Build GRCh38
rs767184882 Chr.12: 67658806-67658806 on Build GRCh38
rs954629958 Chr.12: 67658769-67658769 on Build GRCh38
rs777922597 Chr.12: 67658719-67658719 on Build GRCh38
rs761070005 Chr.12: 67658731-67658731 on Build GRCh38
rs772092011 Chr.12: 67658738-67658738 on Build GRCh38
rs751580966 Chr.12: 67658677-67658677 on Build GRCh38
rs376929227 Chr.12: 67658749-67658749 on Build GRCh38
rs202181002 Chr.12: 67658725-67658725 on Build GRCh38
rs772929446 Chr.12: 67658736-67658736 on Build GRCh38
rs757410743 Chr.12: 67658762-67658762 on Build GRCh38

View Full Product Details