Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31839432-31839705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf48
Entrez Gene ID
50854
Gene Name
chromosome 6 open reading frame 48
Gene Aliases
D6S57, G8
UniGene
Hs.640836
Gene Chromosome Location
Chr.6: 31834915-31839766 on Build GRCh38
Transcripts
NM_001040437.2 NM_001040438.2 NM_001287482.1 NM_001287483.1 NM_001287484.1 NM_001287485.1 NM_001287486.1 NM_001287487.1 NM_001287488.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
223072 c.218C>T p.P73L Chr.6: 31839653-31839653 on Build GRCh38
164507 c.220G>A p.E74K Chr.6: 31839655-31839655 on Build GRCh38
3347020 c.210A>G p.G70G Chr.6: 31839645-31839645 on Build GRCh38
6616492 c.227_228insG p.*76* Chr.6: 31839662-31839663 on Build GRCh38
279446 c.210delA p.M72fs*>4 Chr.6: 31839645-31839645 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1002249961 Chr.6: 31839607-31839607 on Build GRCh38
rs767687857 Chr.6: 31839525-31839526 on Build GRCh38
rs770340681 Chr.6: 31839594-31839594 on Build GRCh38
rs747887431 Chr.6: 31839613-31839613 on Build GRCh38
rs777209897 Chr.6: 31839629-31839629 on Build GRCh38
rs539040262 Chr.6: 31839597-31839597 on Build GRCh38
rs550988160 Chr.6: 31839598-31839598 on Build GRCh38
rs756648264 Chr.6: 31839553-31839554 on Build GRCh38
rs201386093 Chr.6: 31839604-31839604 on Build GRCh38
rs766992140 Chr.6: 31839585-31839585 on Build GRCh38
rs769695769 Chr.6: 31839550-31839550 on Build GRCh38
rs375708709 Chr.6: 31839567-31839567 on Build GRCh38
rs779910185 Chr.6: 31839603-31839603 on Build GRCh38
rs746724674 Chr.6: 31839663-31839663 on Build GRCh38
rs567821977 Chr.6: 31839491-31839491 on Build GRCh38
rs754185843 Chr.6: 31839565-31839565 on Build GRCh38
rs566412576 Chr.6: 31839644-31839644 on Build GRCh38
rs752981971 Chr.6: 31839638-31839638 on Build GRCh38
rs754226294 Chr.6: 31839518-31839518 on Build GRCh38
rs754310022 Chr.6: 31839588-31839588 on Build GRCh38
rs760006801 Chr.6: 31839633-31839633 on Build GRCh38
rs775302260 Chr.6: 31839558-31839558 on Build GRCh38
rs755334986 Chr.6: 31839589-31839589 on Build GRCh38
rs750903223 Chr.6: 31839496-31839496 on Build GRCh38
rs200881992 Chr.6: 31839560-31839560 on Build GRCh38
rs755321008 Chr.6: 31839524-31839524 on Build GRCh38
rs372106776 Chr.6: 31839667-31839667 on Build GRCh38
rs763274522 Chr.6: 31839653-31839653 on Build GRCh38
rs751653805 Chr.6: 31839661-31839661 on Build GRCh38
rs772578932 Chr.6: 31839576-31839576 on Build GRCh38
rs745770085 Chr.6: 31839544-31839544 on Build GRCh38
rs755433878 Chr.6: 31839577-31839577 on Build GRCh38
rs751560288 Chr.6: 31839600-31839600 on Build GRCh38
rs796720032 Chr.6: 31839489-31839489 on Build GRCh38
rs368175561 Chr.6: 31839665-31839665 on Build GRCh38
rs923320235 Chr.6: 31839509-31839510 on Build GRCh38
rs530736278 Chr.6: 31839489-31839492 on Build GRCh38
rs766670549 Chr.6: 31839557-31839558 on Build GRCh38
rs766904002 Chr.6: 31839511-31839511 on Build GRCh38
rs55877173 Chr.6: 31839583-31839583 on Build GRCh38
rs140463292 Chr.6: 31839599-31839599 on Build GRCh38
rs1019065375 Chr.6: 31839487-31839487 on Build GRCh38
rs771575282 Chr.6: 31839616-31839616 on Build GRCh38
rs979673042 Chr.6: 31839488-31839488 on Build GRCh38
rs756461726 Chr.6: 31839499-31839499 on Build GRCh38
rs767008749 Chr.6: 31839573-31839573 on Build GRCh38
rs781021800 Chr.6: 31839541-31839541 on Build GRCh38
rs757607905 Chr.6: 31839534-31839535 on Build GRCh38
rs772847811 Chr.6: 31839614-31839614 on Build GRCh38
rs374923326 Chr.6: 31839495-31839495 on Build GRCh38
rs568902564 Chr.6: 31839581-31839581 on Build GRCh38
rs758434206 Chr.6: 31839535-31839535 on Build GRCh38
rs75582401 Chr.6: 31839566-31839566 on Build GRCh38
rs779261240 Chr.6: 31839529-31839529 on Build GRCh38
rs765588209 Chr.6: 31839636-31839636 on Build GRCh38
rs999670538 Chr.6: 31839465-31839465 on Build GRCh38
rs748522998 Chr.6: 31839489-31839489 on Build GRCh38
rs764216514 Chr.6: 31839654-31839654 on Build GRCh38
rs760251972 Chr.6: 31839615-31839615 on Build GRCh38
rs761261649 Chr.6: 31839569-31839569 on Build GRCh38
rs56062145 Chr.6: 31839528-31839528 on Build GRCh38
rs1034710515 Chr.6: 31839618-31839618 on Build GRCh38
rs748185523 Chr.6: 31839534-31839534 on Build GRCh38
rs777256427 Chr.6: 31839645-31839645 on Build GRCh38

View Full Product Details