Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 31402994-31403231 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRG1
Entrez Gene ID
4733
Gene Name
developmentally regulated GTP binding protein 1
Gene Aliases
NEDD3
UniGene
Hs.115242
Gene Chromosome Location
Chr.22: 31399553-31434186 on Build GRCh38
Transcripts
NM_004147.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5787723 c.197G>T p.R66L Chr.22: 31403059-31403059 on Build GRCh38
7124771 c.318C>G p.I106M Chr.22: 31403180-31403180 on Build GRCh38
6885837 c.249C>T p.N83N Chr.22: 31403111-31403111 on Build GRCh38
6319357 c.167G>T p.G56V Chr.22: 31403029-31403029 on Build GRCh38
4928059 c.178G>T p.A60S Chr.22: 31403040-31403040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769880372 Chr.22: 31403194-31403194 on Build GRCh38
rs778269909 Chr.22: 31403117-31403117 on Build GRCh38
rs17851833 Chr.22: 31403134-31403134 on Build GRCh38
rs879164478 Chr.22: 31403121-31403121 on Build GRCh38
rs759985026 Chr.22: 31403193-31403193 on Build GRCh38
rs944608187 Chr.22: 31403201-31403201 on Build GRCh38
rs758402604 Chr.22: 31403115-31403115 on Build GRCh38
rs779461309 Chr.22: 31403084-31403084 on Build GRCh38
rs747445902 Chr.22: 31403178-31403178 on Build GRCh38
rs910493129 Chr.22: 31403143-31403143 on Build GRCh38
rs777140947 Chr.22: 31403192-31403192 on Build GRCh38
rs549007416 Chr.22: 31403141-31403141 on Build GRCh38
rs771428238 Chr.22: 31403183-31403183 on Build GRCh38
rs755599886 Chr.22: 31403061-31403061 on Build GRCh38
rs748938612 Chr.22: 31403110-31403110 on Build GRCh38
rs768472446 Chr.22: 31403112-31403112 on Build GRCh38
rs752737251 Chr.22: 31403054-31403054 on Build GRCh38

View Full Product Details