Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 112466033-112466258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTLA
Entrez Gene ID
151888
Gene Name
B and T lymphocyte associated
Gene Aliases
BTLA1, CD272
UniGene
Hs.445162
Gene Chromosome Location
Chr.3: 112458790-112499756 on Build GRCh38
Transcripts
NM_001085357.1 NM_181780.3 XM_011512447.2 XM_017005748.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6263233 c.800C>T p.P267L Chr.3: 112466178-112466178 on Build GRCh38
6544660 c.788C>A p.S263Y Chr.3: 112466190-112466190 on Build GRCh38
3585395 c.851C>T p.S284F Chr.3: 112466127-112466127 on Build GRCh38
7302408 c.848C>T p.A283V Chr.3: 112466130-112466130 on Build GRCh38
1484439 c.726T>G p.V242V Chr.3: 112466252-112466252 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766848690 Chr.3: 112466144-112466144 on Build GRCh38
rs770019001 Chr.3: 112466085-112466085 on Build GRCh38
rs767706334 Chr.3: 112466110-112466110 on Build GRCh38
rs769062837 Chr.3: 112466088-112466088 on Build GRCh38
rs35030966 Chr.3: 112466087-112466088 on Build GRCh38
rs2171513 Chr.3: 112466080-112466080 on Build GRCh38
rs749433757 Chr.3: 112466215-112466215 on Build GRCh38
rs138132778 Chr.3: 112466169-112466169 on Build GRCh38
rs116209341 Chr.3: 112466090-112466090 on Build GRCh38
rs758247730 Chr.3: 112466179-112466179 on Build GRCh38
rs113604811 Chr.3: 112466177-112466177 on Build GRCh38
rs771037485 Chr.3: 112466188-112466188 on Build GRCh38
rs768795122 Chr.3: 112466224-112466224 on Build GRCh38
rs761121604 Chr.3: 112466123-112466123 on Build GRCh38
rs781267002 Chr.3: 112466195-112466195 on Build GRCh38
rs753413000 Chr.3: 112466161-112466161 on Build GRCh38
rs771707369 Chr.3: 112466059-112466059 on Build GRCh38
rs1033450737 Chr.3: 112466226-112466226 on Build GRCh38
rs746883407 Chr.3: 112466082-112466082 on Build GRCh38
rs750970562 Chr.3: 112466118-112466118 on Build GRCh38
rs762249928 Chr.3: 112466105-112466105 on Build GRCh38
rs746800681 Chr.3: 112466185-112466185 on Build GRCh38
rs9288952 Chr.3: 112466178-112466178 on Build GRCh38
rs748034910 Chr.3: 112466058-112466058 on Build GRCh38
rs777385531 Chr.3: 112466184-112466184 on Build GRCh38

View Full Product Details