Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 37294850-37295075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA4
Entrez Gene ID
2803
Gene Name
golgin A4
Gene Aliases
CRPF46, GCP2, GOLG, MU-RMS-40.18, p230
UniGene
Hs.344151
Gene Chromosome Location
Chr.3: 37243191-37366879 on Build GRCh38
Transcripts
NM_001172713.1 NM_002078.4 XM_005265069.3 XM_005265070.3 XM_005265071.3 XM_005265072.3 XM_005265073.1 XM_005265074.1 XM_005265075.1 XM_006713110.2 XM_011533598.2 XM_017006184.1 XM_017006185.1 XM_017006186.1 XM_017006187.1 XM_017006188.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3774954 c.670C>G p.L224V Chr.3: 37295066-37295066 on Build GRCh38
6672635 c.583-6delT p.? Chr.3: 37294973-37294973 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs144791036 Chr.3: 37295029-37295029 on Build GRCh38
rs371398817 Chr.3: 37295005-37295005 on Build GRCh38
rs147726248 Chr.3: 37295018-37295018 on Build GRCh38
rs746374684 Chr.3: 37294942-37294942 on Build GRCh38
rs776887458 Chr.3: 37294965-37294965 on Build GRCh38
rs772648656 Chr.3: 37294944-37294944 on Build GRCh38
rs142965770 Chr.3: 37294990-37294990 on Build GRCh38
rs757912281 Chr.3: 37294942-37294942 on Build GRCh38
rs773776488 Chr.3: 37294930-37294930 on Build GRCh38
rs745308692 Chr.3: 37295038-37295038 on Build GRCh38
rs375759795 Chr.3: 37294992-37294992 on Build GRCh38
rs755890979 Chr.3: 37295017-37295017 on Build GRCh38
rs768923352 Chr.3: 37294953-37294953 on Build GRCh38
rs76283087 Chr.3: 37294945-37294945 on Build GRCh38
rs761979187 Chr.3: 37294969-37294969 on Build GRCh38
rs920051427 Chr.3: 37294973-37294973 on Build GRCh38
rs761080488 Chr.3: 37294949-37294949 on Build GRCh38
rs561383963 Chr.3: 37294910-37294910 on Build GRCh38
rs202188013 Chr.3: 37294987-37294987 on Build GRCh38
rs1002809586 Chr.3: 37294896-37294896 on Build GRCh38
rs765351500 Chr.3: 37294975-37294975 on Build GRCh38
rs771738799 Chr.3: 37295045-37295045 on Build GRCh38
rs752946862 Chr.3: 37294882-37294882 on Build GRCh38
rs779679609 Chr.3: 37294944-37294977 on Build GRCh38
rs752666222 Chr.3: 37295015-37295015 on Build GRCh38
rs756927634 Chr.3: 37295025-37295025 on Build GRCh38
rs530320999 Chr.3: 37294926-37294926 on Build GRCh38

View Full Product Details