Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 112154886-112155145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL18
Entrez Gene ID
3606
Gene Name
interleukin 18
Gene Aliases
IGIF, IL-18, IL-1g, IL1F4
UniGene
Hs.83077
Gene Chromosome Location
Chr.11: 112143251-112164117 on Build GRCh38
Transcripts
NM_001243211.1 NM_001562.3 XM_011542805.1 XM_011542806.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3808352 c.48G>A p.M16I Chr.11: 112155006-112155006 on Build GRCh38
1351283 c.66G>A p.T22T Chr.11: 112154988-112154988 on Build GRCh38
3967312 c.65C>T p.T22M Chr.11: 112154989-112154989 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs200077778 Chr.11: 112154988-112154988 on Build GRCh38
rs768151939 Chr.11: 112154962-112154962 on Build GRCh38
rs774621216 Chr.11: 112155041-112155041 on Build GRCh38
rs764661045 Chr.11: 112155102-112155102 on Build GRCh38
rs533003344 Chr.11: 112155111-112155111 on Build GRCh38
rs963670616 Chr.11: 112155064-112155064 on Build GRCh38
rs753178388 Chr.11: 112155084-112155084 on Build GRCh38
rs778247422 Chr.11: 112154925-112154925 on Build GRCh38
rs1005445531 Chr.11: 112154974-112154974 on Build GRCh38
rs1022003947 Chr.11: 112155100-112155100 on Build GRCh38
rs200333982 Chr.11: 112155065-112155065 on Build GRCh38
rs564229852 Chr.11: 112154913-112154913 on Build GRCh38
rs779845094 Chr.11: 112154949-112154949 on Build GRCh38
rs766654584 Chr.11: 112154999-112154999 on Build GRCh38
rs758161059 Chr.11: 112154938-112154938 on Build GRCh38
rs201790534 Chr.11: 112154968-112154968 on Build GRCh38
rs879027850 Chr.11: 112154963-112154964 on Build GRCh38
rs748077794 Chr.11: 112154969-112154969 on Build GRCh38
rs756644856 Chr.11: 112155092-112155092 on Build GRCh38
rs548583841 Chr.11: 112155103-112155103 on Build GRCh38
rs372426039 Chr.11: 112154954-112154954 on Build GRCh38
rs767569150 Chr.11: 112155072-112155072 on Build GRCh38
rs200969913 Chr.11: 112154953-112154953 on Build GRCh38
rs201646620 Chr.11: 112155055-112155055 on Build GRCh38
rs61734549 Chr.11: 112154989-112154989 on Build GRCh38
rs745541879 Chr.11: 112154930-112154930 on Build GRCh38

View Full Product Details