Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133390576-133390849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STKLD1
Entrez Gene ID
169436
Gene Name
serine/threonine kinase like domain containing 1
Gene Aliases
C9orf96, SgK071, Sk521
UniGene
Hs.720195
Gene Chromosome Location
Chr.9: 133376413-133406096 on Build GRCh38
Transcripts
NM_153710.4 NR_103997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1756022 c.537T>C p.N179N Chr.9: 133390750-133390750 on Build GRCh38
3685529 c.468-8G>T p.? Chr.9: 133390673-133390673 on Build GRCh38
1739576 c.572G>A p.R191H Chr.9: 133390785-133390785 on Build GRCh38
3929896 c.496A>T p.I166F Chr.9: 133390709-133390709 on Build GRCh38
4958124 c.557C>T p.A186V Chr.9: 133390770-133390770 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782413996 Chr.9: 133390669-133390669 on Build GRCh38
rs909990153 Chr.9: 133390600-133390600 on Build GRCh38
rs782811199 Chr.9: 133390736-133390736 on Build GRCh38
rs950313486 Chr.9: 133390627-133390627 on Build GRCh38
rs370999988 Chr.9: 133390784-133390784 on Build GRCh38
rs972791671 Chr.9: 133390622-133390622 on Build GRCh38
rs115159388 Chr.9: 133390601-133390601 on Build GRCh38
rs11340207 Chr.9: 133390827-133390827 on Build GRCh38
rs782516525 Chr.9: 133390743-133390743 on Build GRCh38
rs782698733 Chr.9: 133390721-133390721 on Build GRCh38
rs782035275 Chr.9: 133390829-133390829 on Build GRCh38
rs782739788 Chr.9: 133390750-133390750 on Build GRCh38
rs782352613 Chr.9: 133390803-133390803 on Build GRCh38
rs921487463 Chr.9: 133390819-133390819 on Build GRCh38
rs781855875 Chr.9: 133390741-133390741 on Build GRCh38
rs782090906 Chr.9: 133390708-133390708 on Build GRCh38
rs782448197 Chr.9: 133390775-133390775 on Build GRCh38
rs781793324 Chr.9: 133390769-133390769 on Build GRCh38
rs917201267 Chr.9: 133390623-133390623 on Build GRCh38
rs782221543 Chr.9: 133390796-133390796 on Build GRCh38
rs752380119 Chr.9: 133390662-133390662 on Build GRCh38
rs782500796 Chr.9: 133390641-133390641 on Build GRCh38
rs782507810 Chr.9: 133390799-133390799 on Build GRCh38
rs199720626 Chr.9: 133390826-133390826 on Build GRCh38
rs368285833 Chr.9: 133390816-133390816 on Build GRCh38
rs782392501 Chr.9: 133390685-133390685 on Build GRCh38
rs781928660 Chr.9: 133390722-133390722 on Build GRCh38
rs888136405 Chr.9: 133390626-133390626 on Build GRCh38
rs57981115 Chr.9: 133390814-133390814 on Build GRCh38
rs781912295 Chr.9: 133390813-133390813 on Build GRCh38
rs782653973 Chr.9: 133390789-133390789 on Build GRCh38
rs587672490 Chr.9: 133390788-133390788 on Build GRCh38
rs782500873 Chr.9: 133390704-133390706 on Build GRCh38
rs781791626 Chr.9: 133390638-133390639 on Build GRCh38
rs587723955 Chr.9: 133390815-133390815 on Build GRCh38
rs782043745 Chr.9: 133390673-133390673 on Build GRCh38
rs374609912 Chr.9: 133390785-133390785 on Build GRCh38
rs374489407 Chr.9: 133390681-133390681 on Build GRCh38
rs782201633 Chr.9: 133390650-133390650 on Build GRCh38
rs1010398243 Chr.9: 133390763-133390763 on Build GRCh38
rs782605786 Chr.9: 133390794-133390794 on Build GRCh38
rs781949252 Chr.9: 133390700-133390700 on Build GRCh38
rs950562137 Chr.9: 133390701-133390701 on Build GRCh38
rs782149698 Chr.9: 133390725-133390725 on Build GRCh38

View Full Product Details