Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 8452785-8453058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMT44
Entrez Gene ID
152992
Gene Name
tRNA methyltransferase 44 homolog (S. cerevisiae)
Gene Aliases
C4orf23, METTL19, TRM44
UniGene
Hs.566191
Gene Chromosome Location
Chr.4: 8440774-8512531 on Build GRCh38
Transcripts
NM_152544.2 XM_005247950.3 XM_011513406.2 XM_011513407.2 XM_011513408.1 XM_011513409.2 XM_011513410.2 XM_017007811.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7302240 c.331A>G p.R111G Chr.4: 8452912-8452912 on Build GRCh38
7302239 c.1054A>G p.R352G Chr.4: 8452912-8452912 on Build GRCh38
5670544 c.1093G>T p.G365* Chr.4: 8452951-8452951 on Build GRCh38
3393019 c.408+8C>G p.? Chr.4: 8452997-8452997 on Build GRCh38
5060513 c.342C>A p.A114A Chr.4: 8452923-8452923 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992687965 Chr.4: 8452987-8452987 on Build GRCh38
rs376434493 Chr.4: 8452834-8452834 on Build GRCh38
rs758715252 Chr.4: 8452850-8452850 on Build GRCh38
rs768320433 Chr.4: 8452979-8452979 on Build GRCh38
rs1000389066 Chr.4: 8452827-8452827 on Build GRCh38
rs1009710921 Chr.4: 8452879-8452879 on Build GRCh38
rs940057123 Chr.4: 8452974-8452974 on Build GRCh38
rs542469488 Chr.4: 8452833-8452833 on Build GRCh38
rs759818756 Chr.4: 8452903-8452903 on Build GRCh38
rs190439742 Chr.4: 8452887-8452887 on Build GRCh38
rs765695023 Chr.4: 8452906-8452906 on Build GRCh38
rs756940464 Chr.4: 8452914-8452914 on Build GRCh38
rs923688313 Chr.4: 8452942-8452942 on Build GRCh38
rs948657326 Chr.4: 8452989-8452989 on Build GRCh38
rs1044479378 Chr.4: 8452817-8452817 on Build GRCh38
rs751817339 Chr.4: 8452931-8452931 on Build GRCh38
rs146248190 Chr.4: 8452944-8452944 on Build GRCh38
rs184427543 Chr.4: 8452880-8452880 on Build GRCh38
rs975687502 Chr.4: 8452911-8452911 on Build GRCh38
rs749313029 Chr.4: 8452977-8452977 on Build GRCh38
rs749954830 Chr.4: 8452932-8452932 on Build GRCh38
rs1880024 Chr.4: 8452912-8452912 on Build GRCh38
rs185883969 Chr.4: 8452998-8452998 on Build GRCh38
rs750905347 Chr.4: 8452819-8452819 on Build GRCh38
rs781302387 Chr.4: 8453036-8453036 on Build GRCh38
rs193087059 Chr.4: 8452968-8452968 on Build GRCh38
rs778919160 Chr.4: 8453029-8453029 on Build GRCh38
rs779858859 Chr.4: 8452959-8452959 on Build GRCh38
rs917071135 Chr.4: 8452988-8452988 on Build GRCh38
rs952750721 Chr.4: 8452925-8452925 on Build GRCh38
rs778810925 Chr.4: 8452995-8452995 on Build GRCh38

View Full Product Details