Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 69174952-69175177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNB1
Entrez Gene ID
891
Gene Name
cyclin B1
Gene Aliases
CCNB
UniGene
Hs.23960
Gene Chromosome Location
Chr.5: 69167010-69178245 on Build GRCh38
Transcripts
NM_031966.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1069681 c.862A>C p.K288Q Chr.5: 69175033-69175033 on Build GRCh38
1660010 c.865A>G p.I289V Chr.5: 69175036-69175036 on Build GRCh38
4430540 c.922A>T p.R308* Chr.5: 69175093-69175093 on Build GRCh38
1069682 c.864G>T p.K288N Chr.5: 69175035-69175035 on Build GRCh38
5576516 c.790C>T p.P264S Chr.5: 69174961-69174961 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150507468 Chr.5: 69175137-69175137 on Build GRCh38
rs370267052 Chr.5: 69175106-69175106 on Build GRCh38
rs777089233 Chr.5: 69175133-69175133 on Build GRCh38
rs961709268 Chr.5: 69175013-69175013 on Build GRCh38
rs746574794 Chr.5: 69175071-69175071 on Build GRCh38
rs138386021 Chr.5: 69175091-69175091 on Build GRCh38
rs760150623 Chr.5: 69175142-69175143 on Build GRCh38
rs141625969 Chr.5: 69175002-69175002 on Build GRCh38
rs761932194 Chr.5: 69175109-69175109 on Build GRCh38
rs753686417 Chr.5: 69174995-69174995 on Build GRCh38
rs11552410 Chr.5: 69175066-69175066 on Build GRCh38
rs776088624 Chr.5: 69175090-69175090 on Build GRCh38
rs770400177 Chr.5: 69175082-69175082 on Build GRCh38
rs993169524 Chr.5: 69175077-69175077 on Build GRCh38
rs375679998 Chr.5: 69175098-69175098 on Build GRCh38
rs778859090 Chr.5: 69175023-69175023 on Build GRCh38
rs1622483 Chr.5: 69175039-69175039 on Build GRCh38
rs904927260 Chr.5: 69175052-69175052 on Build GRCh38
rs527340846 Chr.5: 69175120-69175120 on Build GRCh38
rs150496399 Chr.5: 69175053-69175053 on Build GRCh38
rs185368107 Chr.5: 69175127-69175127 on Build GRCh38
rs898864873 Chr.5: 69175076-69175076 on Build GRCh38
rs201317423 Chr.5: 69175140-69175140 on Build GRCh38
rs770630440 Chr.5: 69175075-69175075 on Build GRCh38
rs529932616 Chr.5: 69175080-69175080 on Build GRCh38
rs777373420 Chr.5: 69175067-69175067 on Build GRCh38
rs758362213 Chr.5: 69175132-69175132 on Build GRCh38
rs752219518 Chr.5: 69175115-69175116 on Build GRCh38
rs765784179 Chr.5: 69175153-69175154 on Build GRCh38
rs752538536 Chr.5: 69175129-69175129 on Build GRCh38
rs762102951 Chr.5: 69174978-69174978 on Build GRCh38
rs984129710 Chr.5: 69174987-69174987 on Build GRCh38
rs548382997 Chr.5: 69175081-69175081 on Build GRCh38
rs868589726 Chr.5: 69175070-69175070 on Build GRCh38
rs745535905 Chr.5: 69175143-69175143 on Build GRCh38
rs759378255 Chr.5: 69175124-69175124 on Build GRCh38
rs138556581 Chr.5: 69175139-69175139 on Build GRCh38
rs949182365 Chr.5: 69174993-69174993 on Build GRCh38

View Full Product Details