Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27463761-27464008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA2
Entrez Gene ID
1135
Gene Name
cholinergic receptor nicotinic alpha 2 subunit
Gene Aliases
-
UniGene
Hs.57718
Gene Chromosome Location
Chr.8: 27459761-27479296 on Build GRCh38
Transcripts
NM_000742.3 NM_001282455.1 XM_005273397.2 XM_006716282.1 XM_011544388.1 XM_011544389.1 XM_017013003.1 XM_017013004.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1201118 c.506G>A p.G169D Chr.8: 27463937-27463937 on Build GRCh38
226627 c.678G>A p.W226* Chr.8: 27463765-27463765 on Build GRCh38
5595726 c.669G>T p.K223N Chr.8: 27463774-27463774 on Build GRCh38
1201119 c.529G>A p.A177T Chr.8: 27463914-27463914 on Build GRCh38
4999299 c.462G>C p.E154D Chr.8: 27463981-27463981 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546670055 Chr.8: 27463799-27463799 on Build GRCh38
rs772750196 Chr.8: 27463917-27463917 on Build GRCh38
rs750100538 Chr.8: 27463846-27463846 on Build GRCh38
rs992212068 Chr.8: 27463877-27463877 on Build GRCh38
rs960814491 Chr.8: 27463838-27463838 on Build GRCh38
rs1013860481 Chr.8: 27463981-27463981 on Build GRCh38
rs756730967 Chr.8: 27463971-27463971 on Build GRCh38
rs748234276 Chr.8: 27463784-27463784 on Build GRCh38
rs768575325 Chr.8: 27463900-27463900 on Build GRCh38
rs752751788 Chr.8: 27463875-27463875 on Build GRCh38
rs749496919 Chr.8: 27463885-27463885 on Build GRCh38
rs148009259 Chr.8: 27463867-27463867 on Build GRCh38
rs1022647938 Chr.8: 27463816-27463816 on Build GRCh38
rs751893575 Chr.8: 27463951-27463951 on Build GRCh38
rs756175616 Chr.8: 27463880-27463880 on Build GRCh38
rs779984445 Chr.8: 27463889-27463889 on Build GRCh38
rs768536727 Chr.8: 27463820-27463820 on Build GRCh38
rs766697348 Chr.8: 27463862-27463862 on Build GRCh38
rs775471233 Chr.8: 27463807-27463807 on Build GRCh38
rs754976731 Chr.8: 27463894-27463895 on Build GRCh38
rs762644191 Chr.8: 27463847-27463847 on Build GRCh38
rs761237311 Chr.8: 27463834-27463834 on Build GRCh38
rs771350209 Chr.8: 27463888-27463888 on Build GRCh38
rs796052299 Chr.8: 27463853-27463853 on Build GRCh38
rs535699325 Chr.8: 27463954-27463954 on Build GRCh38
rs760660239 Chr.8: 27463809-27463809 on Build GRCh38
rs376045534 Chr.8: 27463961-27463961 on Build GRCh38
rs200523775 Chr.8: 27463792-27463792 on Build GRCh38
rs1009464639 Chr.8: 27463808-27463808 on Build GRCh38
rs373358886 Chr.8: 27463866-27463866 on Build GRCh38
rs778807798 Chr.8: 27463916-27463916 on Build GRCh38
rs141662252 Chr.8: 27463884-27463884 on Build GRCh38
rs201551533 Chr.8: 27463903-27463903 on Build GRCh38
rs746797491 Chr.8: 27463897-27463897 on Build GRCh38
rs781562475 Chr.8: 27463870-27463870 on Build GRCh38
rs769273715 Chr.8: 27463916-27463916 on Build GRCh38
rs1026442070 Chr.8: 27463918-27463918 on Build GRCh38
rs764830161 Chr.8: 27463843-27463843 on Build GRCh38
rs903839863 Chr.8: 27463817-27463817 on Build GRCh38
rs529695608 Chr.8: 27463796-27463796 on Build GRCh38
rs753162068 Chr.8: 27463970-27463970 on Build GRCh38
rs796052298 Chr.8: 27463914-27463914 on Build GRCh38
rs192887571 Chr.8: 27463906-27463906 on Build GRCh38
rs766076722 Chr.8: 27463940-27463940 on Build GRCh38
rs201279312 Chr.8: 27463791-27463791 on Build GRCh38
rs796052308 Chr.8: 27463885-27463886 on Build GRCh38
rs377660408 Chr.8: 27463790-27463790 on Build GRCh38
rs762588688 Chr.8: 27463921-27463921 on Build GRCh38
rs776152904 Chr.8: 27463825-27463825 on Build GRCh38
rs370425843 Chr.8: 27463828-27463828 on Build GRCh38

View Full Product Details