Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3784484-3784749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPB
Entrez Gene ID
1059
Gene Name
centromere protein B
Gene Aliases
-
UniGene
Hs.516855
Gene Chromosome Location
Chr.20: 3783851-3786690 on Build GRCh38
Transcripts
NM_001810.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7076359 c.1792C>G p.Q598E Chr.20: 3784692-3784692 on Build GRCh38
5414881 c.1743G>C p.V581V Chr.20: 3784741-3784741 on Build GRCh38
3713308 c.1769C>T p.A590V Chr.20: 3784715-3784715 on Build GRCh38
3546275 c.1784T>C p.L595P Chr.20: 3784700-3784700 on Build GRCh38
6626480 c.1759G>A p.A587T Chr.20: 3784725-3784725 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747014298 Chr.20: 3784715-3784715 on Build GRCh38
rs975214960 Chr.20: 3784540-3784540 on Build GRCh38
rs750078348 Chr.20: 3784688-3784688 on Build GRCh38
rs930727352 Chr.20: 3784658-3784658 on Build GRCh38
rs901746698 Chr.20: 3784610-3784610 on Build GRCh38
rs1018748997 Chr.20: 3784605-3784605 on Build GRCh38
rs940895082 Chr.20: 3784505-3784505 on Build GRCh38
rs757341919 Chr.20: 3784725-3784725 on Build GRCh38
rs551453910 Chr.20: 3784533-3784533 on Build GRCh38
rs992340964 Chr.20: 3784512-3784512 on Build GRCh38
rs6052076 Chr.20: 3784531-3784531 on Build GRCh38
rs767027838 Chr.20: 3784685-3784685 on Build GRCh38
rs986471746 Chr.20: 3784526-3784526 on Build GRCh38
rs369514352 Chr.20: 3784681-3784681 on Build GRCh38
rs188528517 Chr.20: 3784657-3784657 on Build GRCh38
rs770114131 Chr.20: 3784647-3784647 on Build GRCh38
rs544897093 Chr.20: 3784520-3784520 on Build GRCh38
rs61734519 Chr.20: 3784726-3784726 on Build GRCh38
rs776044169 Chr.20: 3784660-3784660 on Build GRCh38
rs756008400 Chr.20: 3784696-3784696 on Build GRCh38
rs912443731 Chr.20: 3784530-3784530 on Build GRCh38
rs763676260 Chr.20: 3784664-3784664 on Build GRCh38
rs763438322 Chr.20: 3784663-3784663 on Build GRCh38
rs935749493 Chr.20: 3784527-3784527 on Build GRCh38
rs773666307 Chr.20: 3784671-3784671 on Build GRCh38
rs996074233 Chr.20: 3784636-3784636 on Build GRCh38
rs754812338 Chr.20: 3784707-3784707 on Build GRCh38
rs918138541 Chr.20: 3784515-3784515 on Build GRCh38
rs566661302 Chr.20: 3784603-3784603 on Build GRCh38
rs376597377 Chr.20: 3784714-3784714 on Build GRCh38
rs753881816 Chr.20: 3784706-3784706 on Build GRCh38
rs766268613 Chr.20: 3784697-3784697 on Build GRCh38
rs1010453627 Chr.20: 3784544-3784544 on Build GRCh38
rs562991696 Chr.20: 3784521-3784521 on Build GRCh38
rs375448007 Chr.20: 3784633-3784633 on Build GRCh38
rs777409066 Chr.20: 3784615-3784615 on Build GRCh38

View Full Product Details