Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 179678563-179678792 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CANX
Entrez Gene ID
821
Gene Name
calnexin
Gene Aliases
CNX, IP90, P90
UniGene
Hs.567968
Gene Chromosome Location
Chr.5: 179678631-179731641 on Build GRCh38
Transcripts
XM_011534665.2 NM_001164444.1 XM_017009726.1

Gene Symbol
CBY3
Entrez Gene ID
646019
Gene Name
chibby family member 3
Gene Aliases
-
UniGene
Hs.535767
Gene Chromosome Location
Chr.5: 179677969-179681830 on Build GRCh38
Transcripts
XM_011534665.2 NM_001164444.1 XM_017009726.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5991776 c.616_622delCCCGAGG p.P206fs*1 Chr.5: 179678690-179678696 on Build GRCh38
4159809 c.638C>G p.A213G Chr.5: 179678674-179678674 on Build GRCh38
6221731 c.611G>T p.R204L Chr.5: 179678701-179678701 on Build GRCh38
4987896 c.719A>C p.D240A Chr.5: 179678593-179678593 on Build GRCh38
6581176 c.627C>T p.I209I Chr.5: 179678685-179678685 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs114542354 Chr.5: 179678700-179678700 on Build GRCh38
rs550624803 Chr.5: 179678661-179678661 on Build GRCh38
rs909011310 Chr.5: 179678581-179678581 on Build GRCh38
rs904667876 Chr.5: 179678655-179678655 on Build GRCh38
rs538946054 Chr.5: 179678585-179678585 on Build GRCh38
rs555166898 Chr.5: 179678656-179678656 on Build GRCh38
rs772438184 Chr.5: 179678664-179678664 on Build GRCh38
rs1024678750 Chr.5: 179678671-179678671 on Build GRCh38
rs569095817 Chr.5: 179678615-179678615 on Build GRCh38
rs765055756 Chr.5: 179678690-179678690 on Build GRCh38
rs751081425 Chr.5: 179678701-179678701 on Build GRCh38
rs750194507 Chr.5: 179678694-179678694 on Build GRCh38
rs201900739 Chr.5: 179678675-179678675 on Build GRCh38
rs868724108 Chr.5: 179678725-179678725 on Build GRCh38
rs780929318 Chr.5: 179678613-179678613 on Build GRCh38
rs762743798 Chr.5: 179678643-179678643 on Build GRCh38
rs1051816691 Chr.5: 179678747-179678747 on Build GRCh38
rs558988617 Chr.5: 179678696-179678696 on Build GRCh38
rs562797198 Chr.5: 179678709-179678709 on Build GRCh38
rs760632581 Chr.5: 179678665-179678665 on Build GRCh38
rs946286208 Chr.5: 179678625-179678625 on Build GRCh38
rs774855235 Chr.5: 179678649-179678649 on Build GRCh38
rs199692218 Chr.5: 179678593-179678593 on Build GRCh38
rs78613293 Chr.5: 179678674-179678674 on Build GRCh38
rs892039992 Chr.5: 179678623-179678623 on Build GRCh38
rs894515132 Chr.5: 179678598-179678598 on Build GRCh38
rs1043287274 Chr.5: 179678731-179678731 on Build GRCh38
rs772013802 Chr.5: 179678753-179678753 on Build GRCh38
rs768382885 Chr.5: 179678603-179678603 on Build GRCh38
rs200177340 Chr.5: 179678698-179678698 on Build GRCh38
rs1043464911 Chr.5: 179678724-179678724 on Build GRCh38
rs1016612059 Chr.5: 179678676-179678676 on Build GRCh38
rs749688064 Chr.5: 179678614-179678614 on Build GRCh38
rs963099567 Chr.5: 179678672-179678672 on Build GRCh38
rs746451968 Chr.5: 179678601-179678601 on Build GRCh38
rs546644235 Chr.5: 179678726-179678726 on Build GRCh38
rs947373672 Chr.5: 179678604-179678604 on Build GRCh38
rs779126754 Chr.5: 179678722-179678722 on Build GRCh38
rs1013253632 Chr.5: 179678667-179678667 on Build GRCh38
rs751423576 Chr.5: 179678589-179678589 on Build GRCh38
rs748417764 Chr.5: 179678735-179678735 on Build GRCh38
rs903254283 Chr.5: 179678631-179678631 on Build GRCh38
rs530023226 Chr.5: 179678730-179678730 on Build GRCh38
rs780839309 Chr.5: 179678729-179678729 on Build GRCh38
rs982365257 Chr.5: 179678583-179678583 on Build GRCh38
rs868115996 Chr.5: 179678616-179678616 on Build GRCh38
rs927808035 Chr.5: 179678682-179678682 on Build GRCh38
rs774832399 Chr.5: 179678593-179678594 on Build GRCh38
rs369839116 Chr.5: 179678679-179678679 on Build GRCh38
rs975006182 Chr.5: 179678686-179678686 on Build GRCh38
rs567422903 Chr.5: 179678642-179678642 on Build GRCh38
rs757288930 Chr.5: 179678657-179678657 on Build GRCh38
rs1033413513 Chr.5: 179678658-179678658 on Build GRCh38
rs922032487 Chr.5: 179678596-179678596 on Build GRCh38

View Full Product Details