Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31333534-31333803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSG1
Entrez Gene ID
1828
Gene Name
desmoglein 1
Gene Aliases
CDHF4, DG1, DSG, EPKHE, EPKHIA, PPKS1, SPPK1
UniGene
Hs.2633
Gene Chromosome Location
Chr.18: 31318089-31359246 on Build GRCh38
Transcripts
NM_001942.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
395669 c.692G>T p.G231V Chr.18: 31333596-31333596 on Build GRCh38
5578443 c.725G>A p.R242Q Chr.18: 31333629-31333629 on Build GRCh38
2153245 c.724C>T p.R242* Chr.18: 31333628-31333628 on Build GRCh38
4130948 c.732C>T p.G244G Chr.18: 31333636-31333636 on Build GRCh38
2809138 c.799C>A p.P267T Chr.18: 31333703-31333703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558523078 Chr.18: 31333748-31333748 on Build GRCh38
rs779510702 Chr.18: 31333761-31333761 on Build GRCh38
rs759636098 Chr.18: 31333735-31333735 on Build GRCh38
rs371339794 Chr.18: 31333592-31333592 on Build GRCh38
rs201719724 Chr.18: 31333596-31333596 on Build GRCh38
rs753393332 Chr.18: 31333766-31333766 on Build GRCh38
rs748648648 Chr.18: 31333656-31333656 on Build GRCh38
rs751417742 Chr.18: 31333586-31333586 on Build GRCh38
rs753832211 Chr.18: 31333572-31333572 on Build GRCh38
rs754548405 Chr.18: 31333767-31333767 on Build GRCh38
rs934792343 Chr.18: 31333672-31333672 on Build GRCh38
rs772482149 Chr.18: 31333657-31333657 on Build GRCh38
rs781133396 Chr.18: 31333594-31333594 on Build GRCh38
rs568609861 Chr.18: 31333628-31333628 on Build GRCh38
rs766472053 Chr.18: 31333710-31333710 on Build GRCh38
rs748694136 Chr.18: 31333635-31333635 on Build GRCh38
rs199911116 Chr.18: 31333715-31333715 on Build GRCh38
rs750142736 Chr.18: 31333757-31333757 on Build GRCh38
rs761832336 Chr.18: 31333749-31333749 on Build GRCh38
rs534042917 Chr.18: 31333708-31333708 on Build GRCh38
rs764078467 Chr.18: 31333579-31333579 on Build GRCh38
rs774113449 Chr.18: 31333577-31333578 on Build GRCh38
rs758393297 Chr.18: 31333577-31333577 on Build GRCh38
rs145045312 Chr.18: 31333638-31333638 on Build GRCh38
rs146862520 Chr.18: 31333671-31333671 on Build GRCh38
rs751685288 Chr.18: 31333620-31333620 on Build GRCh38
rs140546771 Chr.18: 31333684-31333684 on Build GRCh38
rs767458166 Chr.18: 31333751-31333751 on Build GRCh38
rs993351359 Chr.18: 31333559-31333559 on Build GRCh38
rs12967407 Chr.18: 31333636-31333636 on Build GRCh38
rs375634680 Chr.18: 31333760-31333760 on Build GRCh38
rs142797072 Chr.18: 31333607-31333607 on Build GRCh38
rs535358304 Chr.18: 31333595-31333595 on Build GRCh38
rs56032720 Chr.18: 31333747-31333747 on Build GRCh38
rs146538145 Chr.18: 31333633-31333633 on Build GRCh38
rs761494614 Chr.18: 31333581-31333589 on Build GRCh38
rs987513671 Chr.18: 31333613-31333613 on Build GRCh38

View Full Product Details