Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 16200596-16200865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT15
Entrez Gene ID
117248
Gene Name
polypeptide N-acetylgalactosaminyltransferase 15
Gene Aliases
GALNACT15, GALNT13, GALNT7, GALNTL2, PIH5, pp-GalNAc-T15
UniGene
Hs.411308
Gene Chromosome Location
Chr.3: 16174322-16245190 on Build GRCh38
Transcripts
NM_001319051.1 NM_054110.4 XM_005264852.4 XM_017005682.1 XM_017005683.1 XM_017005684.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4750392 c.796G>T p.A266S Chr.3: 16200708-16200708 on Build GRCh38
1420338 c.750delG p.V252fs*1 Chr.3: 16200662-16200662 on Build GRCh38
3589516 c.847G>A p.D283N Chr.3: 16200759-16200759 on Build GRCh38
4411660 c.859G>T p.E287* Chr.3: 16200771-16200771 on Build GRCh38
1669996 c.811G>C p.A271P Chr.3: 16200723-16200723 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs929791127 Chr.3: 16200803-16200803 on Build GRCh38
rs555860390 Chr.3: 16200764-16200764 on Build GRCh38
rs143956414 Chr.3: 16200686-16200686 on Build GRCh38
rs773048639 Chr.3: 16200618-16200618 on Build GRCh38
rs766617201 Chr.3: 16200747-16200747 on Build GRCh38
rs745413922 Chr.3: 16200711-16200711 on Build GRCh38
rs750493713 Chr.3: 16200744-16200744 on Build GRCh38
rs866277477 Chr.3: 16200673-16200673 on Build GRCh38
rs772728197 Chr.3: 16200829-16200830 on Build GRCh38
rs777114326 Chr.3: 16200732-16200732 on Build GRCh38
rs1000227925 Chr.3: 16200649-16200649 on Build GRCh38
rs780458853 Chr.3: 16200685-16200685 on Build GRCh38
rs771537146 Chr.3: 16200770-16200770 on Build GRCh38
rs761198489 Chr.3: 16200699-16200722 on Build GRCh38
rs776794357 Chr.3: 16200813-16200813 on Build GRCh38
rs764363495 Chr.3: 16200725-16200725 on Build GRCh38
rs370015694 Chr.3: 16200638-16200638 on Build GRCh38
rs534853378 Chr.3: 16200621-16200621 on Build GRCh38
rs765779157 Chr.3: 16200743-16200743 on Build GRCh38
rs150729824 Chr.3: 16200810-16200810 on Build GRCh38
rs1043548093 Chr.3: 16200646-16200646 on Build GRCh38
rs369377608 Chr.3: 16200822-16200822 on Build GRCh38
rs764567497 Chr.3: 16200641-16200641 on Build GRCh38
rs760414905 Chr.3: 16200724-16200724 on Build GRCh38
rs758577456 Chr.3: 16200746-16200746 on Build GRCh38
rs749165221 Chr.3: 16200693-16200693 on Build GRCh38
rs200564483 Chr.3: 16200795-16200795 on Build GRCh38
rs769536107 Chr.3: 16200725-16200726 on Build GRCh38
rs200118171 Chr.3: 16200619-16200619 on Build GRCh38
rs903713667 Chr.3: 16200648-16200648 on Build GRCh38
rs557520950 Chr.3: 16200623-16200623 on Build GRCh38
rs1011794669 Chr.3: 16200630-16200630 on Build GRCh38
rs767157320 Chr.3: 16200634-16200634 on Build GRCh38
rs7638396 Chr.3: 16200722-16200722 on Build GRCh38
rs541983333 Chr.3: 16200762-16200762 on Build GRCh38
rs774877715 Chr.3: 16200824-16200824 on Build GRCh38
rs202129269 Chr.3: 16200636-16200636 on Build GRCh38
rs944355966 Chr.3: 16200629-16200629 on Build GRCh38
rs918393109 Chr.3: 16200800-16200800 on Build GRCh38
rs778842843 Chr.3: 16200766-16200766 on Build GRCh38
rs1024949558 Chr.3: 16200696-16200696 on Build GRCh38
rs374048513 Chr.3: 16200749-16200749 on Build GRCh38
rs758630696 Chr.3: 16200682-16200682 on Build GRCh38
rs372040056 Chr.3: 16200712-16200712 on Build GRCh38
rs759127078 Chr.3: 16200628-16200628 on Build GRCh38
rs201490177 Chr.3: 16200737-16200737 on Build GRCh38
rs773779764 Chr.3: 16200709-16200709 on Build GRCh38
rs757851207 Chr.3: 16200642-16200642 on Build GRCh38
rs573371305 Chr.3: 16200666-16200666 on Build GRCh38
rs755282595 Chr.3: 16200750-16200750 on Build GRCh38
rs536900672 Chr.3: 16200707-16200707 on Build GRCh38
rs778845492 Chr.3: 16200658-16200658 on Build GRCh38
rs777370991 Chr.3: 16200752-16200752 on Build GRCh38

View Full Product Details