Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 39144874-39145127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSRNP1
Entrez Gene ID
64651
Gene Name
cysteine and serine rich nuclear protein 1
Gene Aliases
AXUD1, CSRNP-1, FAM130B, TAIP-3, URAX1
UniGene
Hs.370950
Gene Chromosome Location
Chr.3: 39141851-39154602 on Build GRCh38
Transcripts
NM_001320559.1 NM_001320560.1 NM_033027.3 XM_017007048.1 XM_017007049.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5650110 c.439_441delGAG p.E147delE Chr.3: 39145021-39145023 on Build GRCh38
1202520 c.365G>A p.R122H Chr.3: 39145097-39145097 on Build GRCh38
5449867 c.361C>T p.R121C Chr.3: 39145101-39145101 on Build GRCh38
6405995 c.432G>T p.L144F Chr.3: 39145030-39145030 on Build GRCh38
1422917 c.407G>A p.R136Q Chr.3: 39145055-39145055 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562210829 Chr.3: 39145047-39145047 on Build GRCh38
rs757921966 Chr.3: 39145061-39145061 on Build GRCh38
rs866090111 Chr.3: 39145076-39145076 on Build GRCh38
rs757603631 Chr.3: 39145051-39145051 on Build GRCh38
rs149009984 Chr.3: 39145055-39145055 on Build GRCh38
rs765195891 Chr.3: 39145024-39145024 on Build GRCh38
rs977078871 Chr.3: 39144918-39144918 on Build GRCh38
rs776893085 Chr.3: 39145076-39145076 on Build GRCh38
rs201759916 Chr.3: 39145075-39145075 on Build GRCh38
rs779714471 Chr.3: 39145062-39145062 on Build GRCh38
rs977043195 Chr.3: 39144967-39144967 on Build GRCh38
rs746438028 Chr.3: 39145056-39145056 on Build GRCh38
rs145992009 Chr.3: 39145034-39145034 on Build GRCh38
rs138067710 Chr.3: 39145023-39145023 on Build GRCh38
rs746601332 Chr.3: 39145063-39145063 on Build GRCh38
rs760720188 Chr.3: 39145100-39145100 on Build GRCh38
rs776520739 Chr.3: 39145020-39145020 on Build GRCh38
rs150622788 Chr.3: 39145035-39145035 on Build GRCh38
rs776192489 Chr.3: 39145073-39145073 on Build GRCh38
rs771777455 Chr.3: 39144974-39144974 on Build GRCh38
rs773078092 Chr.3: 39145087-39145087 on Build GRCh38
rs200798768 Chr.3: 39145097-39145097 on Build GRCh38
rs768268943 Chr.3: 39145010-39145010 on Build GRCh38
rs369362214 Chr.3: 39145101-39145101 on Build GRCh38
rs769542490 Chr.3: 39145074-39145074 on Build GRCh38
rs756378294 Chr.3: 39144949-39144949 on Build GRCh38
rs749723210 Chr.3: 39144958-39144958 on Build GRCh38
rs967733230 Chr.3: 39144935-39144935 on Build GRCh38
rs536835334 Chr.3: 39144973-39144973 on Build GRCh38
rs771491132 Chr.3: 39144962-39144962 on Build GRCh38
rs201210294 Chr.3: 39144927-39144927 on Build GRCh38
rs762883711 Chr.3: 39145088-39145088 on Build GRCh38
rs775266749 Chr.3: 39144978-39144978 on Build GRCh38
rs778207977 Chr.3: 39144953-39144953 on Build GRCh38
rs139691127 Chr.3: 39145040-39145040 on Build GRCh38
rs967142256 Chr.3: 39144902-39144902 on Build GRCh38
rs374212431 Chr.3: 39145041-39145041 on Build GRCh38
rs372648220 Chr.3: 39145050-39145050 on Build GRCh38
rs772116804 Chr.3: 39145067-39145071 on Build GRCh38
rs764320842 Chr.3: 39145048-39145048 on Build GRCh38
rs779613659 Chr.3: 39144965-39144965 on Build GRCh38
rs372028126 Chr.3: 39145098-39145098 on Build GRCh38
rs562295707 Chr.3: 39144903-39144903 on Build GRCh38
rs768372949 Chr.3: 39145064-39145064 on Build GRCh38
rs368112233 Chr.3: 39144979-39144979 on Build GRCh38

View Full Product Details