Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27469558-27469817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA2
Entrez Gene ID
1135
Gene Name
cholinergic receptor nicotinic alpha 2 subunit
Gene Aliases
-
UniGene
Hs.57718
Gene Chromosome Location
Chr.8: 27459761-27479296 on Build GRCh38
Transcripts
NM_000742.3 NM_001282455.1 XM_005273397.2 XM_006716282.1 XM_011544388.1 XM_011544389.1 XM_017013003.1 XM_017013004.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4996940 c.242C>T p.S81L Chr.8: 27469813-27469813 on Build GRCh38
1456392 c.277G>A p.A93T Chr.8: 27469778-27469778 on Build GRCh38
6628692 c.289G>A p.D97N Chr.8: 27469766-27469766 on Build GRCh38
6862770 c.260G>A p.R87H Chr.8: 27469795-27469795 on Build GRCh38
5746855 c.294+1G>A p.? Chr.8: 27469760-27469760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368211446 Chr.8: 27469742-27469742 on Build GRCh38
rs1034238568 Chr.8: 27469735-27469735 on Build GRCh38
rs748420222 Chr.8: 27469741-27469741 on Build GRCh38
rs982512969 Chr.8: 27469619-27469619 on Build GRCh38
rs749766964 Chr.8: 27469746-27469746 on Build GRCh38
rs116459673 Chr.8: 27469661-27469661 on Build GRCh38
rs903912119 Chr.8: 27469725-27469725 on Build GRCh38
rs537861239 Chr.8: 27469640-27469640 on Build GRCh38
rs1050980684 Chr.8: 27469669-27469669 on Build GRCh38
rs771491769 Chr.8: 27469755-27469755 on Build GRCh38
rs201944711 Chr.8: 27469779-27469779 on Build GRCh38
rs376078367 Chr.8: 27469733-27469733 on Build GRCh38
rs749083602 Chr.8: 27469705-27469705 on Build GRCh38
rs772244275 Chr.8: 27469759-27469759 on Build GRCh38
rs751049729 Chr.8: 27469778-27469778 on Build GRCh38
rs371576634 Chr.8: 27469728-27469728 on Build GRCh38
rs374208928 Chr.8: 27469711-27469711 on Build GRCh38
rs371304320 Chr.8: 27469665-27469665 on Build GRCh38
rs552736727 Chr.8: 27469664-27469664 on Build GRCh38
rs190255317 Chr.8: 27469706-27469706 on Build GRCh38
rs995570511 Chr.8: 27469702-27469702 on Build GRCh38
rs552944382 Chr.8: 27469607-27469607 on Build GRCh38
rs775714882 Chr.8: 27469766-27469766 on Build GRCh38
rs561008438 Chr.8: 27469767-27469767 on Build GRCh38
rs752371632 Chr.8: 27469785-27469785 on Build GRCh38
rs367739392 Chr.8: 27469586-27469586 on Build GRCh38
rs992440390 Chr.8: 27469604-27469604 on Build GRCh38
rs544902258 Chr.8: 27469689-27469689 on Build GRCh38
rs754622688 Chr.8: 27469737-27469737 on Build GRCh38
rs755757952 Chr.8: 27469786-27469786 on Build GRCh38
rs149615415 Chr.8: 27469769-27469769 on Build GRCh38
rs778048020 Chr.8: 27469745-27469745 on Build GRCh38
rs774398407 Chr.8: 27469756-27469756 on Build GRCh38
rs765040068 Chr.8: 27469724-27469724 on Build GRCh38
rs778191765 Chr.8: 27469796-27469796 on Build GRCh38
rs975942177 Chr.8: 27469618-27469618 on Build GRCh38
rs147191290 Chr.8: 27469612-27469612 on Build GRCh38
rs199963019 Chr.8: 27469734-27469734 on Build GRCh38
rs185419685 Chr.8: 27469645-27469645 on Build GRCh38
rs532212433 Chr.8: 27469795-27469795 on Build GRCh38
rs780994616 Chr.8: 27469738-27469738 on Build GRCh38
rs976289821 Chr.8: 27469754-27469754 on Build GRCh38
rs892447964 Chr.8: 27469677-27469677 on Build GRCh38
rs776346092 Chr.8: 27469720-27469720 on Build GRCh38
rs766434161 Chr.8: 27469777-27469777 on Build GRCh38
rs568484025 Chr.8: 27469648-27469648 on Build GRCh38
rs745958182 Chr.8: 27469757-27469757 on Build GRCh38
rs916888102 Chr.8: 27469605-27469605 on Build GRCh38
rs767229210 Chr.8: 27469781-27469781 on Build GRCh38
rs945418539 Chr.8: 27469681-27469681 on Build GRCh38

View Full Product Details