Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48434810-48435043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC51
Entrez Gene ID
79714
Gene Name
coiled-coil domain containing 51
Gene Aliases
-
UniGene
Hs.187657
Gene Chromosome Location
Chr.3: 48432170-48446652 on Build GRCh38
Transcripts
NM_001256964.1 NM_001256965.2 NM_001256966.2 NM_001256967.2 NM_001256968.2 NM_001256969.2 NM_024661.4 XM_011534113.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3594913 c.250G>A p.D84N Chr.3: 48434879-48434879 on Build GRCh38
1045815 c.110G>A p.C37Y Chr.3: 48435019-48435019 on Build GRCh38
4118198 c.205C>T p.H69Y Chr.3: 48434924-48434924 on Build GRCh38
7411115 c.237C>T p.A79A Chr.3: 48434892-48434892 on Build GRCh38
6302512 c.172C>A p.H58N Chr.3: 48434957-48434957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762424234 Chr.3: 48435014-48435014 on Build GRCh38
rs779069056 Chr.3: 48434987-48434987 on Build GRCh38
rs762557010 Chr.3: 48434947-48434947 on Build GRCh38
rs751257115 Chr.3: 48434953-48434953 on Build GRCh38
rs749944612 Chr.3: 48434946-48434946 on Build GRCh38
rs771455628 Chr.3: 48434904-48434904 on Build GRCh38
rs376560818 Chr.3: 48434948-48434948 on Build GRCh38
rs776307065 Chr.3: 48435005-48435005 on Build GRCh38
rs772327618 Chr.3: 48434994-48434994 on Build GRCh38
rs771037375 Chr.3: 48434902-48434902 on Build GRCh38
rs780139053 Chr.3: 48434954-48434956 on Build GRCh38
rs11553587 Chr.3: 48434930-48434930 on Build GRCh38
rs61746705 Chr.3: 48434997-48434997 on Build GRCh38
rs746028126 Chr.3: 48434906-48434906 on Build GRCh38
rs368305792 Chr.3: 48434850-48434850 on Build GRCh38
rs754355881 Chr.3: 48434967-48434967 on Build GRCh38
rs529283500 Chr.3: 48434847-48434847 on Build GRCh38
rs758064467 Chr.3: 48434861-48434861 on Build GRCh38
rs778062593 Chr.3: 48434958-48434958 on Build GRCh38
rs370066515 Chr.3: 48434979-48434979 on Build GRCh38
rs749795179 Chr.3: 48434896-48434896 on Build GRCh38
rs1045900789 Chr.3: 48434931-48434931 on Build GRCh38
rs376034975 Chr.3: 48434857-48434857 on Build GRCh38
rs764750440 Chr.3: 48434943-48434943 on Build GRCh38
rs13062723 Chr.3: 48435021-48435021 on Build GRCh38
rs759522565 Chr.3: 48435019-48435020 on Build GRCh38
rs551032462 Chr.3: 48434942-48434942 on Build GRCh38
rs113958192 Chr.3: 48434851-48434851 on Build GRCh38
rs374404532 Chr.3: 48434988-48434988 on Build GRCh38
rs748540384 Chr.3: 48434871-48434871 on Build GRCh38
rs776627148 Chr.3: 48434911-48434911 on Build GRCh38
rs769758588 Chr.3: 48435008-48435008 on Build GRCh38
rs761745752 Chr.3: 48435007-48435007 on Build GRCh38
rs200464374 Chr.3: 48434842-48434842 on Build GRCh38
rs756467640 Chr.3: 48434954-48434954 on Build GRCh38
rs781471796 Chr.3: 48434866-48434866 on Build GRCh38
rs948860119 Chr.3: 48434928-48434928 on Build GRCh38
rs760498722 Chr.3: 48434909-48434909 on Build GRCh38
rs371682973 Chr.3: 48434908-48434908 on Build GRCh38
rs775436647 Chr.3: 48434843-48434843 on Build GRCh38
rs775136508 Chr.3: 48435011-48435011 on Build GRCh38
rs774630527 Chr.3: 48434905-48434905 on Build GRCh38
rs61740637 Chr.3: 48434887-48434887 on Build GRCh38
rs756527266 Chr.3: 48434886-48434886 on Build GRCh38
rs199770348 Chr.3: 48434996-48434996 on Build GRCh38
rs764115835 Chr.3: 48434849-48434849 on Build GRCh38
rs1011939669 Chr.3: 48434970-48434970 on Build GRCh38
rs1459248 Chr.3: 48434983-48434983 on Build GRCh38

View Full Product Details