Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 42510777-42511050 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM1
Entrez Gene ID
634
Gene Name
carcinoembryonic antigen related cell adhesion molecule 1
Gene Aliases
BGP, BGP1, BGPI
UniGene
Hs.512682
Gene Chromosome Location
Chr.19: 42507306-42528509 on Build GRCh38
Transcripts
NM_001024912.2 NM_001184813.1 NM_001184815.1 NM_001184816.1 NM_001205344.1 NM_001712.4 NR_073179.1 NR_073180.1

Gene Symbol
LIPE-AS1
Entrez Gene ID
100996307
Gene Name
LIPE antisense RNA 1
Gene Aliases
-
UniGene
Hs.466840
Gene Chromosome Location
Chr.19: 42397148-42652355 on Build GRCh38
Transcripts
NM_001024912.2 NM_001184813.1 NM_001184815.1 NM_001184816.1 NM_001205344.1 NM_001712.4 NR_073179.1 NR_073180.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3773050 c.1432C>A p.Q478K Chr.19: 42510918-42510918 on Build GRCh38
4283690 c.1266+5G>A p.? Chr.19: 42510884-42510884 on Build GRCh38
3287867 c.1461+5G>A p.? Chr.19: 42510884-42510884 on Build GRCh38
3773051 c.1237C>A p.Q413K Chr.19: 42510918-42510918 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs931062942 Chr.19: 42510826-42510829 on Build GRCh38
rs1032334940 Chr.19: 42510987-42510987 on Build GRCh38
rs775565476 Chr.19: 42510904-42510904 on Build GRCh38
rs953203853 Chr.19: 42510996-42510996 on Build GRCh38
rs750773439 Chr.19: 42510949-42510949 on Build GRCh38
rs374692844 Chr.19: 42510877-42510877 on Build GRCh38
rs554896669 Chr.19: 42510958-42510958 on Build GRCh38
rs996886055 Chr.19: 42510964-42510964 on Build GRCh38
rs146856369 Chr.19: 42510897-42510897 on Build GRCh38
rs760661371 Chr.19: 42510905-42510905 on Build GRCh38
rs759785203 Chr.19: 42510898-42510898 on Build GRCh38
rs747541385 Chr.19: 42510864-42510864 on Build GRCh38
rs749172222 Chr.19: 42510879-42510879 on Build GRCh38
rs764731994 Chr.19: 42510939-42510939 on Build GRCh38
rs769611863 Chr.19: 42510850-42510850 on Build GRCh38
rs751232014 Chr.19: 42510845-42510845 on Build GRCh38
rs780877846 Chr.19: 42510855-42510855 on Build GRCh38
rs769784160 Chr.19: 42510876-42510876 on Build GRCh38
rs879031825 Chr.19: 42511013-42511013 on Build GRCh38
rs772215589 Chr.19: 42510902-42510902 on Build GRCh38
rs371121061 Chr.19: 42510852-42510852 on Build GRCh38
rs775213687 Chr.19: 42510904-42510905 on Build GRCh38
rs886949157 Chr.19: 42510861-42510861 on Build GRCh38
rs762284761 Chr.19: 42510948-42510948 on Build GRCh38
rs376219357 Chr.19: 42510831-42510831 on Build GRCh38
rs1023375449 Chr.19: 42510812-42510812 on Build GRCh38
rs868154375 Chr.19: 42510918-42510918 on Build GRCh38
rs368995051 Chr.19: 42510839-42510839 on Build GRCh38
rs199595296 Chr.19: 42510896-42510896 on Build GRCh38
rs368457248 Chr.19: 42510945-42510945 on Build GRCh38

View Full Product Details