Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27461460-27461733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA2
Entrez Gene ID
1135
Gene Name
cholinergic receptor nicotinic alpha 2 subunit
Gene Aliases
-
UniGene
Hs.57718
Gene Chromosome Location
Chr.8: 27459761-27479296 on Build GRCh38
Transcripts
NM_000742.3 NM_001282455.1 XM_005273397.2 XM_006716282.1 XM_011544388.1 XM_011544389.1 XM_017013003.1 XM_017013004.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1755677 c.1543G>T p.G515W Chr.8: 27461676-27461676 on Build GRCh38
1098733 c.1555C>A p.L519I Chr.8: 27461664-27461664 on Build GRCh38
7165700 c.1513C>T p.L505F Chr.8: 27461706-27461706 on Build GRCh38
3647808 c.1580G>A p.G527E Chr.8: 27461639-27461639 on Build GRCh38
4397625 c.1567C>T p.P523S Chr.8: 27461652-27461652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974292330 Chr.8: 27461582-27461582 on Build GRCh38
rs775950981 Chr.8: 27461694-27461695 on Build GRCh38
rs777504286 Chr.8: 27461596-27461596 on Build GRCh38
rs528528743 Chr.8: 27461691-27461691 on Build GRCh38
rs746176483 Chr.8: 27461710-27461710 on Build GRCh38
rs934551659 Chr.8: 27461606-27461606 on Build GRCh38
rs908646142 Chr.8: 27461554-27461554 on Build GRCh38
rs199993564 Chr.8: 27461650-27461650 on Build GRCh38
rs776005376 Chr.8: 27461619-27461619 on Build GRCh38
rs759372688 Chr.8: 27461488-27461488 on Build GRCh38
rs780560860 Chr.8: 27461574-27461574 on Build GRCh38
rs753434826 Chr.8: 27461672-27461672 on Build GRCh38
rs772407561 Chr.8: 27461618-27461618 on Build GRCh38
rs770505681 Chr.8: 27461621-27461621 on Build GRCh38
rs748966513 Chr.8: 27461599-27461599 on Build GRCh38
rs147882344 Chr.8: 27461521-27461521 on Build GRCh38
rs762018858 Chr.8: 27461645-27461645 on Build GRCh38
rs560295005 Chr.8: 27461519-27461519 on Build GRCh38
rs760680104 Chr.8: 27461621-27461621 on Build GRCh38
rs978024767 Chr.8: 27461583-27461583 on Build GRCh38
rs755631456 Chr.8: 27461667-27461667 on Build GRCh38
rs796052306 Chr.8: 27461688-27461688 on Build GRCh38
rs75394008 Chr.8: 27461575-27461575 on Build GRCh38
rs768734385 Chr.8: 27461626-27461626 on Build GRCh38
rs149142237 Chr.8: 27461689-27461689 on Build GRCh38
rs368132207 Chr.8: 27461627-27461627 on Build GRCh38
rs777412930 Chr.8: 27461668-27461668 on Build GRCh38
rs375304755 Chr.8: 27461625-27461625 on Build GRCh38
rs145238683 Chr.8: 27461651-27461651 on Build GRCh38
rs964926183 Chr.8: 27461517-27461517 on Build GRCh38
rs555088381 Chr.8: 27461497-27461497 on Build GRCh38
rs1012797739 Chr.8: 27461703-27461703 on Build GRCh38
rs186509115 Chr.8: 27461600-27461600 on Build GRCh38
rs779112876 Chr.8: 27461615-27461615 on Build GRCh38
rs906989445 Chr.8: 27461677-27461677 on Build GRCh38
rs746289570 Chr.8: 27461616-27461616 on Build GRCh38
rs767320056 Chr.8: 27461654-27461654 on Build GRCh38
rs141508075 Chr.8: 27461592-27461592 on Build GRCh38
rs752689106 Chr.8: 27461662-27461662 on Build GRCh38
rs572033587 Chr.8: 27461518-27461518 on Build GRCh38
rs981468883 Chr.8: 27461534-27461534 on Build GRCh38
rs577770461 Chr.8: 27461525-27461525 on Build GRCh38
rs1040135347 Chr.8: 27461644-27461644 on Build GRCh38
rs534403222 Chr.8: 27461520-27461520 on Build GRCh38
rs1016411801 Chr.8: 27461506-27461506 on Build GRCh38

View Full Product Details