Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 119118796-119119061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMXL1
Entrez Gene ID
1657
Gene Name
Dmx like 1
Gene Aliases
-
UniGene
Hs.181042
Gene Chromosome Location
Chr.5: 119071002-119250170 on Build GRCh38
Transcripts
NM_001290321.2 NM_001290322.2 NM_005509.5 XM_005271909.4 XM_005271910.4 XM_005271912.2 XM_011543212.2 XM_011543213.2 XM_011543214.2 XM_011543215.2 XM_017009141.1 XM_017009142.1 XM_017009143.1 XM_017009144.1 XM_017009145.1 XM_017009146.1 XM_017009147.1 XM_017009148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3134415 c.833T>C p.L278P Chr.5: 119118904-119118904 on Build GRCh38
3134417 c.910G>T p.E304* Chr.5: 119118981-119118981 on Build GRCh38
7417546 c.854A>T p.H285L Chr.5: 119118925-119118925 on Build GRCh38
6168463 c.868A>G p.I290V Chr.5: 119118939-119118939 on Build GRCh38
5829999 c.756T>C p.C252C Chr.5: 119118827-119118827 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746370261 Chr.5: 119119012-119119012 on Build GRCh38
rs375271214 Chr.5: 119119025-119119025 on Build GRCh38
rs763705818 Chr.5: 119118824-119118824 on Build GRCh38
rs749067891 Chr.5: 119118978-119118978 on Build GRCh38
rs59649653 Chr.5: 119118901-119118901 on Build GRCh38
rs776356918 Chr.5: 119118994-119118994 on Build GRCh38
rs889672368 Chr.5: 119118862-119118862 on Build GRCh38
rs779854170 Chr.5: 119118857-119118857 on Build GRCh38
rs776127486 Chr.5: 119118895-119118895 on Build GRCh38
rs376075158 Chr.5: 119118831-119118831 on Build GRCh38
rs770281161 Chr.5: 119119021-119119021 on Build GRCh38
rs776982814 Chr.5: 119118907-119118907 on Build GRCh38
rs780110326 Chr.5: 119118867-119118868 on Build GRCh38
rs765552138 Chr.5: 119118911-119118911 on Build GRCh38
rs780958245 Chr.5: 119118833-119118833 on Build GRCh38
rs191440572 Chr.5: 119119030-119119030 on Build GRCh38
rs756696696 Chr.5: 119118934-119118934 on Build GRCh38
rs762711155 Chr.5: 119118918-119118918 on Build GRCh38
rs140972662 Chr.5: 119118920-119118920 on Build GRCh38
rs767000856 Chr.5: 119118948-119118948 on Build GRCh38
rs770938719 Chr.5: 119119001-119119001 on Build GRCh38
rs985097366 Chr.5: 119119022-119119022 on Build GRCh38
rs1047389401 Chr.5: 119118931-119118931 on Build GRCh38
rs560633408 Chr.5: 119118921-119118921 on Build GRCh38
rs886155994 Chr.5: 119118985-119118985 on Build GRCh38
rs372998467 Chr.5: 119118908-119118908 on Build GRCh38
rs981990565 Chr.5: 119118851-119118851 on Build GRCh38
rs373143457 Chr.5: 119119002-119119002 on Build GRCh38
rs200933738 Chr.5: 119118904-119118904 on Build GRCh38
rs748374686 Chr.5: 119118873-119118873 on Build GRCh38
rs777942863 Chr.5: 119118992-119118992 on Build GRCh38
rs761325204 Chr.5: 119119028-119119028 on Build GRCh38
rs745721380 Chr.5: 119118839-119118839 on Build GRCh38
rs747393980 Chr.5: 119118876-119118876 on Build GRCh38
rs779836749 Chr.5: 119118976-119118976 on Build GRCh38
rs367822456 Chr.5: 119118898-119118898 on Build GRCh38
rs771511327 Chr.5: 119118905-119118905 on Build GRCh38
rs200673180 Chr.5: 119118875-119118875 on Build GRCh38
rs549335030 Chr.5: 119119032-119119032 on Build GRCh38
rs758685188 Chr.5: 119118984-119118984 on Build GRCh38
rs775934966 Chr.5: 119118916-119118916 on Build GRCh38
rs755880455 Chr.5: 119118850-119118850 on Build GRCh38
rs750355441 Chr.5: 119118957-119118957 on Build GRCh38
rs781291695 Chr.5: 119119004-119119004 on Build GRCh38
rs376204152 Chr.5: 119118973-119118973 on Build GRCh38
rs751059947 Chr.5: 119118829-119118829 on Build GRCh38

View Full Product Details