Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 31260447-31260672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHD3
Entrez Gene ID
30845
Gene Name
EH domain containing 3
Gene Aliases
PAST3
UniGene
Hs.368808
Gene Chromosome Location
Chr.2: 31234014-31269451 on Build GRCh38
Transcripts
NM_014600.2 XM_011532806.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6010067 c.526G>C p.E176Q Chr.2: 31260533-31260533 on Build GRCh38
3581080 c.579A>T p.K193N Chr.2: 31260586-31260586 on Build GRCh38
4838553 c.664G>A p.D222N Chr.2: 31260671-31260671 on Build GRCh38
1020112 c.643C>T p.R215* Chr.2: 31260650-31260650 on Build GRCh38
4619184 c.503G>C p.G168A Chr.2: 31260510-31260510 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775725614 Chr.2: 31260500-31260500 on Build GRCh38
rs771048215 Chr.2: 31260474-31260474 on Build GRCh38
rs112609329 Chr.2: 31260652-31260652 on Build GRCh38
rs200027155 Chr.2: 31260505-31260505 on Build GRCh38
rs754381951 Chr.2: 31260558-31260558 on Build GRCh38
rs900182342 Chr.2: 31260580-31260580 on Build GRCh38
rs778315422 Chr.2: 31260471-31260471 on Build GRCh38
rs371507212 Chr.2: 31260587-31260587 on Build GRCh38
rs745950962 Chr.2: 31260589-31260589 on Build GRCh38
rs768625383 Chr.2: 31260631-31260631 on Build GRCh38
rs778088572 Chr.2: 31260557-31260557 on Build GRCh38
rs1048803980 Chr.2: 31260651-31260651 on Build GRCh38
rs146103643 Chr.2: 31260610-31260610 on Build GRCh38
rs776537856 Chr.2: 31260636-31260636 on Build GRCh38
rs375784165 Chr.2: 31260600-31260600 on Build GRCh38
rs772124528 Chr.2: 31260599-31260599 on Build GRCh38
rs764883628 Chr.2: 31260526-31260526 on Build GRCh38
rs139542048 Chr.2: 31260522-31260522 on Build GRCh38
rs567062495 Chr.2: 31260504-31260504 on Build GRCh38
rs773132739 Chr.2: 31260640-31260640 on Build GRCh38
rs769628934 Chr.2: 31260638-31260638 on Build GRCh38
rs556019532 Chr.2: 31260544-31260544 on Build GRCh38
rs747225097 Chr.2: 31260623-31260623 on Build GRCh38
rs766849329 Chr.2: 31260483-31260483 on Build GRCh38
rs557753544 Chr.2: 31260637-31260637 on Build GRCh38
rs751708799 Chr.2: 31260485-31260485 on Build GRCh38
rs755067433 Chr.2: 31260488-31260489 on Build GRCh38
rs749920764 Chr.2: 31260527-31260527 on Build GRCh38
rs575875306 Chr.2: 31260549-31260549 on Build GRCh38
rs370390199 Chr.2: 31260489-31260489 on Build GRCh38
rs1057418700 Chr.2: 31260532-31260532 on Build GRCh38
rs778825027 Chr.2: 31260488-31260488 on Build GRCh38
rs772207764 Chr.2: 31260496-31260496 on Build GRCh38
rs751219079 Chr.2: 31260650-31260650 on Build GRCh38
rs776880592 Chr.2: 31260649-31260649 on Build GRCh38
rs537831838 Chr.2: 31260574-31260574 on Build GRCh38
rs762432989 Chr.2: 31260648-31260648 on Build GRCh38
rs146384685 Chr.2: 31260511-31260511 on Build GRCh38

View Full Product Details