Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 34944032-34944285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM47B
Entrez Gene ID
170062
Gene Name
family with sequence similarity 47 member B
Gene Aliases
-
UniGene
Hs.197801
Gene Chromosome Location
Chr.X: 34942796-34944917 on Build GRCh38
Transcripts
NM_152631.2

SNP Detail

SNP ID Chromosome Location
rs757560148 Chr.X: 34944123-34944123 on Build GRCh38
rs148652922 Chr.X: 34944069-34944069 on Build GRCh38
rs761100549 Chr.X: 34944220-34944220 on Build GRCh38
rs761377350 Chr.X: 34944173-34944173 on Build GRCh38
rs772899404 Chr.X: 34944192-34944192 on Build GRCh38
rs139388040 Chr.X: 34944172-34944172 on Build GRCh38
rs767804467 Chr.X: 34944210-34944210 on Build GRCh38
rs771799168 Chr.X: 34944145-34944145 on Build GRCh38
rs777738560 Chr.X: 34944103-34944103 on Build GRCh38
rs756181314 Chr.X: 34944098-34944098 on Build GRCh38
rs144167325 Chr.X: 34944200-34944200 on Build GRCh38
rs758160204 Chr.X: 34944247-34944247 on Build GRCh38
rs749655129 Chr.X: 34944157-34944157 on Build GRCh38
rs146264202 Chr.X: 34944087-34944087 on Build GRCh38
rs753184544 Chr.X: 34944093-34944093 on Build GRCh38
rs112769272 Chr.X: 34944086-34944086 on Build GRCh38
rs367947257 Chr.X: 34944235-34944235 on Build GRCh38
rs769422864 Chr.X: 34944180-34944180 on Build GRCh38
rs768151864 Chr.X: 34944170-34944170 on Build GRCh38
rs776214625 Chr.X: 34944071-34944071 on Build GRCh38
rs759914400 Chr.X: 34944198-34944198 on Build GRCh38
rs200405744 Chr.X: 34944127-34944127 on Build GRCh38
rs779997194 Chr.X: 34944152-34944152 on Build GRCh38
rs143391263 Chr.X: 34944083-34944083 on Build GRCh38
rs1031464198 Chr.X: 34944161-34944161 on Build GRCh38
rs186976890 Chr.X: 34944165-34944165 on Build GRCh38
rs761466788 Chr.X: 34944082-34944082 on Build GRCh38
rs762308715 Chr.X: 34944232-34944232 on Build GRCh38
rs191392189 Chr.X: 34944211-34944211 on Build GRCh38
rs149776639 Chr.X: 34944144-34944144 on Build GRCh38
rs151033833 Chr.X: 34944222-34944222 on Build GRCh38
rs1021316758 Chr.X: 34944059-34944059 on Build GRCh38
rs749150601 Chr.X: 34944066-34944066 on Build GRCh38
rs754058778 Chr.X: 34944122-34944122 on Build GRCh38
rs371374386 Chr.X: 34944245-34944245 on Build GRCh38

View Full Product Details