Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15458119-15458392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLQ
Entrez Gene ID
8292
Gene Name
collagen like tail subunit of asymmetric acetylcholinesterase
Gene Aliases
CMS5, EAD
UniGene
Hs.146735
Gene Chromosome Location
Chr.3: 15450133-15521751 on Build GRCh38
Transcripts
NM_005677.3 NM_080538.2 NM_080539.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4513434 c.908C>T p.S303F Chr.3: 15458202-15458202 on Build GRCh38
6633937 c.914G>A p.R305Q Chr.3: 15458196-15458196 on Build GRCh38
5892854 c.823C>T p.P275S Chr.3: 15458287-15458287 on Build GRCh38
6633935 c.944G>A p.R315Q Chr.3: 15458196-15458196 on Build GRCh38
4432220 c.882C>T p.Y294Y Chr.3: 15458228-15458228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771944316 Chr.3: 15458198-15458198 on Build GRCh38
rs1036386101 Chr.3: 15458208-15458208 on Build GRCh38
rs143918191 Chr.3: 15458203-15458203 on Build GRCh38
rs755182558 Chr.3: 15458150-15458150 on Build GRCh38
rs773686063 Chr.3: 15458317-15458317 on Build GRCh38
rs887744990 Chr.3: 15458164-15458164 on Build GRCh38
rs763262836 Chr.3: 15458320-15458320 on Build GRCh38
rs751621776 Chr.3: 15458236-15458236 on Build GRCh38
rs121908924 Chr.3: 15458197-15458197 on Build GRCh38
rs771406985 Chr.3: 15458330-15458330 on Build GRCh38
rs184245561 Chr.3: 15458218-15458218 on Build GRCh38
rs781304702 Chr.3: 15458176-15458176 on Build GRCh38
rs121908922 Chr.3: 15458296-15458296 on Build GRCh38
rs774736540 Chr.3: 15458337-15458337 on Build GRCh38
rs897137684 Chr.3: 15458354-15458354 on Build GRCh38
rs746460292 Chr.3: 15458287-15458287 on Build GRCh38
rs1037134713 Chr.3: 15458344-15458344 on Build GRCh38
rs766821521 Chr.3: 15458231-15458231 on Build GRCh38
rs763209840 Chr.3: 15458228-15458228 on Build GRCh38
rs755020019 Chr.3: 15458251-15458251 on Build GRCh38
rs550979119 Chr.3: 15458202-15458202 on Build GRCh38
rs377766179 Chr.3: 15458356-15458356 on Build GRCh38
rs114896510 Chr.3: 15458305-15458305 on Build GRCh38
rs763518486 Chr.3: 15458226-15458226 on Build GRCh38
rs756396914 Chr.3: 15458282-15458282 on Build GRCh38
rs375903795 Chr.3: 15458149-15458149 on Build GRCh38
rs1013542424 Chr.3: 15458228-15458228 on Build GRCh38
rs374490483 Chr.3: 15458269-15458269 on Build GRCh38
rs886058099 Chr.3: 15458228-15458228 on Build GRCh38
rs767492114 Chr.3: 15458362-15458362 on Build GRCh38
rs370201862 Chr.3: 15458199-15458199 on Build GRCh38
rs1044349532 Chr.3: 15458215-15458215 on Build GRCh38
rs752905204 Chr.3: 15458271-15458271 on Build GRCh38
rs778740405 Chr.3: 15458196-15458196 on Build GRCh38
rs969251829 Chr.3: 15458343-15458343 on Build GRCh38
rs771444359 Chr.3: 15458336-15458337 on Build GRCh38
rs200403785 Chr.3: 15458284-15458284 on Build GRCh38
rs757060689 Chr.3: 15458185-15458185 on Build GRCh38
rs866348012 Chr.3: 15458249-15458249 on Build GRCh38
rs6782980 Chr.3: 15458206-15458206 on Build GRCh38
rs776425306 Chr.3: 15458212-15458212 on Build GRCh38
rs896388650 Chr.3: 15458213-15458213 on Build GRCh38
rs765680430 Chr.3: 15458222-15458222 on Build GRCh38
rs749018849 Chr.3: 15458180-15458180 on Build GRCh38
rs200232229 Chr.3: 15458214-15458214 on Build GRCh38
rs1014062201 Chr.3: 15458323-15458323 on Build GRCh38
rs773636951 Chr.3: 15458215-15458215 on Build GRCh38
rs1055184620 Chr.3: 15458237-15458237 on Build GRCh38
rs778156526 Chr.3: 15458301-15458301 on Build GRCh38
rs758307097 Chr.3: 15458285-15458285 on Build GRCh38

View Full Product Details