Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 87343663-87343932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO31
Entrez Gene ID
79791
Gene Name
F-box protein 31
Gene Aliases
FBX14, FBXO14, Fbx31, MRT45, pp2386
UniGene
Hs.567582
Gene Chromosome Location
Chr.16: 87326987-87392107 on Build GRCh38
Transcripts
NM_001282683.1 NM_024735.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1252266 c.583C>T p.H195Y Chr.16: 87343672-87343672 on Build GRCh38
6359329 c.535G>C p.D179H Chr.16: 87343720-87343720 on Build GRCh38
379120 c.540C>T p.P180P Chr.16: 87343715-87343715 on Build GRCh38
4063422 c.38C>G p.P13R Chr.16: 87343701-87343701 on Build GRCh38
1709492 c.1A>C p.M1L Chr.16: 87343738-87343738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762500727 Chr.16: 87343712-87343712 on Build GRCh38
rs2303764 Chr.16: 87343849-87343849 on Build GRCh38
rs1035628336 Chr.16: 87343686-87343686 on Build GRCh38
rs745740645 Chr.16: 87343714-87343714 on Build GRCh38
rs755198640 Chr.16: 87343745-87343745 on Build GRCh38
rs752940051 Chr.16: 87343760-87343760 on Build GRCh38
rs768282215 Chr.16: 87343804-87343804 on Build GRCh38
rs1034327421 Chr.16: 87343779-87343779 on Build GRCh38
rs895147319 Chr.16: 87343850-87343850 on Build GRCh38
rs200753805 Chr.16: 87343774-87343774 on Build GRCh38
rs528427644 Chr.16: 87343876-87343876 on Build GRCh38
rs571470392 Chr.16: 87343893-87343893 on Build GRCh38
rs759203274 Chr.16: 87343696-87343696 on Build GRCh38
rs1004430631 Chr.16: 87343902-87343902 on Build GRCh38
rs908151177 Chr.16: 87343896-87343896 on Build GRCh38
rs932943433 Chr.16: 87343821-87343821 on Build GRCh38
rs768252174 Chr.16: 87343768-87343769 on Build GRCh38
rs777895872 Chr.16: 87343775-87343775 on Build GRCh38
rs1016752371 Chr.16: 87343757-87343757 on Build GRCh38
rs886608685 Chr.16: 87343703-87343703 on Build GRCh38
rs56412475 Chr.16: 87343819-87343819 on Build GRCh38
rs770821935 Chr.16: 87343787-87343787 on Build GRCh38
rs933519374 Chr.16: 87343844-87343844 on Build GRCh38
rs1003638505 Chr.16: 87343734-87343734 on Build GRCh38
rs1026891848 Chr.16: 87343892-87343892 on Build GRCh38
rs750749073 Chr.16: 87343812-87343812 on Build GRCh38
rs906579944 Chr.16: 87343835-87343835 on Build GRCh38
rs769368972 Chr.16: 87343699-87343699 on Build GRCh38
rs899227761 Chr.16: 87343911-87343911 on Build GRCh38
rs761155642 Chr.16: 87343718-87343718 on Build GRCh38
rs529786750 Chr.16: 87343891-87343891 on Build GRCh38
rs779067828 Chr.16: 87343759-87343759 on Build GRCh38
rs546557486 Chr.16: 87343831-87343831 on Build GRCh38
rs755559407 Chr.16: 87343898-87343898 on Build GRCh38
rs376068374 Chr.16: 87343732-87343732 on Build GRCh38
rs201696130 Chr.16: 87343805-87343805 on Build GRCh38
rs746951686 Chr.16: 87343780-87343780 on Build GRCh38
rs775309853 Chr.16: 87343708-87343708 on Build GRCh38
rs781007792 Chr.16: 87343789-87343789 on Build GRCh38
rs763706020 Chr.16: 87343716-87343716 on Build GRCh38
rs769730662 Chr.16: 87343793-87343793 on Build GRCh38
rs200061135 Chr.16: 87343694-87343694 on Build GRCh38
rs199880188 Chr.16: 87343803-87343803 on Build GRCh38
rs894364206 Chr.16: 87343709-87343709 on Build GRCh38
rs773844606 Chr.16: 87343770-87343772 on Build GRCh38
rs888304971 Chr.16: 87343901-87343901 on Build GRCh38
rs750898027 Chr.16: 87343717-87343717 on Build GRCh38
rs549227763 Chr.16: 87343838-87343838 on Build GRCh38
rs116482570 Chr.16: 87343830-87343830 on Build GRCh38
rs766837896 Chr.16: 87343811-87343811 on Build GRCh38
rs754207207 Chr.16: 87343738-87343738 on Build GRCh38
rs761226298 Chr.16: 87343810-87343810 on Build GRCh38
rs989682593 Chr.16: 87343823-87343823 on Build GRCh38
rs761545273 Chr.16: 87343805-87343805 on Build GRCh38
rs940837842 Chr.16: 87343899-87343899 on Build GRCh38
rs775221347 Chr.16: 87343797-87343797 on Build GRCh38
rs759771821 Chr.16: 87343813-87343813 on Build GRCh38
rs771990978 Chr.16: 87343766-87343767 on Build GRCh38
rs563390690 Chr.16: 87343827-87343827 on Build GRCh38
rs766920382 Chr.16: 87343842-87343842 on Build GRCh38
rs746478573 Chr.16: 87343767-87343768 on Build GRCh38

View Full Product Details