Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101244912-101245185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRP2
Entrez Gene ID
1821
Gene Name
dystrophin related protein 2
Gene Aliases
DRP-2
UniGene
Hs.159291
Gene Chromosome Location
Chr.X: 101219786-101264497 on Build GRCh38
Transcripts
NM_001171184.1 NM_001939.2 XM_017029333.1 XM_017029334.1

SNP Detail

SNP ID Chromosome Location
rs748173294 Chr.X: 101245112-101245112 on Build GRCh38
rs754380035 Chr.X: 101245021-101245021 on Build GRCh38
rs1015411475 Chr.X: 101244961-101244961 on Build GRCh38
rs371129060 Chr.X: 101245000-101245000 on Build GRCh38
rs755815069 Chr.X: 101245075-101245075 on Build GRCh38
rs376356193 Chr.X: 101245033-101245033 on Build GRCh38
rs763647992 Chr.X: 101245012-101245012 on Build GRCh38
rs919689492 Chr.X: 101245086-101245086 on Build GRCh38
rs200410975 Chr.X: 101245093-101245093 on Build GRCh38
rs758017507 Chr.X: 101245008-101245008 on Build GRCh38
rs780921718 Chr.X: 101245048-101245048 on Build GRCh38
rs749562222 Chr.X: 101245097-101245097 on Build GRCh38
rs572955131 Chr.X: 101245118-101245118 on Build GRCh38
rs934634467 Chr.X: 101245058-101245058 on Build GRCh38
rs772672834 Chr.X: 101245149-101245149 on Build GRCh38
rs917570508 Chr.X: 101245122-101245122 on Build GRCh38
rs745686379 Chr.X: 101245055-101245055 on Build GRCh38
rs773543899 Chr.X: 101245128-101245128 on Build GRCh38
rs766989749 Chr.X: 101244972-101244972 on Build GRCh38
rs746573511 Chr.X: 101244993-101244994 on Build GRCh38
rs868805324 Chr.X: 101245053-101245053 on Build GRCh38
rs768976897 Chr.X: 101245098-101245098 on Build GRCh38
rs866655083 Chr.X: 101245017-101245017 on Build GRCh38
rs757248687 Chr.X: 101245043-101245043 on Build GRCh38
rs367680452 Chr.X: 101245102-101245102 on Build GRCh38

View Full Product Details