Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 17639207-17639476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM184B
Entrez Gene ID
27146
Gene Name
family with sequence similarity 184 member B
Gene Aliases
-
UniGene
Hs.744951
Gene Chromosome Location
Chr.4: 17632073-17781512 on Build GRCh38
Transcripts
NM_015688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4482828 c.2644C>G p.R882G Chr.4: 17639272-17639272 on Build GRCh38
1053363 c.2617A>G p.S873G Chr.4: 17639299-17639299 on Build GRCh38
4538525 c.2562G>A p.V854V Chr.4: 17639354-17639354 on Build GRCh38
1428577 c.2585G>A p.R862Q Chr.4: 17639331-17639331 on Build GRCh38
4937371 c.2537A>G p.Q846R Chr.4: 17639379-17639379 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146323716 Chr.4: 17639247-17639247 on Build GRCh38
rs7655264 Chr.4: 17639288-17639288 on Build GRCh38
rs1031000604 Chr.4: 17639279-17639279 on Build GRCh38
rs536602442 Chr.4: 17639418-17639418 on Build GRCh38
rs968565866 Chr.4: 17639270-17639270 on Build GRCh38
rs758159377 Chr.4: 17639367-17639367 on Build GRCh38
rs1055442924 Chr.4: 17639441-17639441 on Build GRCh38
rs894351579 Chr.4: 17639236-17639236 on Build GRCh38
rs780031151 Chr.4: 17639344-17639344 on Build GRCh38
rs780688794 Chr.4: 17639286-17639286 on Build GRCh38
rs999546226 Chr.4: 17639272-17639272 on Build GRCh38
rs929757061 Chr.4: 17639379-17639379 on Build GRCh38
rs200345030 Chr.4: 17639386-17639386 on Build GRCh38
rs189838036 Chr.4: 17639358-17639358 on Build GRCh38
rs554172404 Chr.4: 17639368-17639368 on Build GRCh38
rs374999361 Chr.4: 17639343-17639343 on Build GRCh38
rs1011841120 Chr.4: 17639241-17639241 on Build GRCh38
rs185814242 Chr.4: 17639262-17639262 on Build GRCh38
rs760760224 Chr.4: 17639338-17639338 on Build GRCh38
rs968106032 Chr.4: 17639452-17639452 on Build GRCh38
rs777579561 Chr.4: 17639446-17639446 on Build GRCh38
rs765356866 Chr.4: 17639354-17639354 on Build GRCh38
rs183021758 Chr.4: 17639417-17639417 on Build GRCh38
rs756786483 Chr.4: 17639269-17639269 on Build GRCh38
rs61742285 Chr.4: 17639257-17639257 on Build GRCh38
rs994992108 Chr.4: 17639271-17639271 on Build GRCh38
rs928337633 Chr.4: 17639389-17639389 on Build GRCh38
rs772048329 Chr.4: 17639378-17639378 on Build GRCh38
rs758076895 Chr.4: 17639252-17639252 on Build GRCh38
rs6811887 Chr.4: 17639235-17639235 on Build GRCh38
rs370211318 Chr.4: 17639251-17639251 on Build GRCh38

View Full Product Details