Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 94445604-94445865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCT
Entrez Gene ID
1638
Gene Name
dopachrome tautomerase
Gene Aliases
TRP-2, TYRP2
UniGene
Hs.301865
Gene Chromosome Location
Chr.13: 94437304-94550265 on Build GRCh38
Transcripts
NM_001129889.2 NM_001322182.1 NM_001322183.1 NM_001322184.1 NM_001322185.1 NM_001322186.1 NM_001922.4 XM_011521049.1 XM_017020401.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6253165 c.1251C>A p.T417T Chr.13: 94445746-94445746 on Build GRCh38
4462908 c.1266C>T p.S422S Chr.13: 94445731-94445731 on Build GRCh38
6333388 c.1278+5C>A p.? Chr.13: 94445714-94445714 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs78182872 Chr.13: 94445712-94445712 on Build GRCh38
rs746252950 Chr.13: 94445735-94445735 on Build GRCh38
rs907142945 Chr.13: 94445630-94445630 on Build GRCh38
rs781527340 Chr.13: 94445728-94445728 on Build GRCh38
rs1015937191 Chr.13: 94445724-94445724 on Build GRCh38
rs369615608 Chr.13: 94445772-94445772 on Build GRCh38
rs116452540 Chr.13: 94445642-94445642 on Build GRCh38
rs567872847 Chr.13: 94445643-94445643 on Build GRCh38
rs756096521 Chr.13: 94445681-94445681 on Build GRCh38
rs538783071 Chr.13: 94445749-94445749 on Build GRCh38
rs199649619 Chr.13: 94445750-94445750 on Build GRCh38
rs1034637403 Chr.13: 94445634-94445634 on Build GRCh38
rs747889652 Chr.13: 94445710-94445710 on Build GRCh38
rs961705434 Chr.13: 94445729-94445729 on Build GRCh38
rs770446816 Chr.13: 94445748-94445748 on Build GRCh38
rs1005361244 Chr.13: 94445706-94445706 on Build GRCh38
rs778440473 Chr.13: 94445708-94445708 on Build GRCh38
rs755684 Chr.13: 94445702-94445702 on Build GRCh38
rs766486413 Chr.13: 94445659-94445659 on Build GRCh38
rs751601140 Chr.13: 94445654-94445654 on Build GRCh38
rs767705767 Chr.13: 94445679-94445679 on Build GRCh38
rs754527174 Chr.13: 94445739-94445741 on Build GRCh38
rs774433019 Chr.13: 94445765-94445765 on Build GRCh38
rs898377207 Chr.13: 94445644-94445644 on Build GRCh38
rs764253216 Chr.13: 94445652-94445652 on Build GRCh38
rs566703183 Chr.13: 94445744-94445744 on Build GRCh38
rs530384136 Chr.13: 94445674-94445674 on Build GRCh38
rs750714880 Chr.13: 94445741-94445741 on Build GRCh38
rs751526005 Chr.13: 94445680-94445681 on Build GRCh38
rs762137393 Chr.13: 94445778-94445778 on Build GRCh38
rs753668033 Chr.13: 94445698-94445698 on Build GRCh38
rs372320239 Chr.13: 94445812-94445812 on Build GRCh38
rs912229332 Chr.13: 94445631-94445631 on Build GRCh38
rs766868561 Chr.13: 94445758-94445758 on Build GRCh38
rs769122969 Chr.13: 94445753-94445753 on Build GRCh38
rs376642164 Chr.13: 94445839-94445839 on Build GRCh38
rs756329617 Chr.13: 94445743-94445743 on Build GRCh38
rs750331477 Chr.13: 94445680-94445680 on Build GRCh38
rs537857314 Chr.13: 94445824-94445824 on Build GRCh38
rs779777615 Chr.13: 94445696-94445696 on Build GRCh38
rs564237351 Chr.13: 94445783-94445783 on Build GRCh38
rs1011758838 Chr.13: 94445752-94445752 on Build GRCh38
rs911381193 Chr.13: 94445805-94445806 on Build GRCh38
rs963710123 Chr.13: 94445835-94445835 on Build GRCh38
rs775694325 Chr.13: 94445799-94445799 on Build GRCh38
rs41300594 Chr.13: 94445667-94445667 on Build GRCh38
rs1024347010 Chr.13: 94445711-94445711 on Build GRCh38

View Full Product Details