Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 582125-582350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSG
Entrez Gene ID
682
Gene Name
basigin (Ok blood group)
Gene Aliases
5F7, CD147, EMMPRIN, OK, TCSF
UniGene
Hs.501293
Gene Chromosome Location
Chr.19: 571277-583493 on Build GRCh38
Transcripts
NM_001322243.1 NM_001728.3 NM_198589.2 NM_198590.2 NM_198591.2 XM_017027173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3540522 c.1070-1G>A p.? Chr.19: 582305-582305 on Build GRCh38
1680445 c.1076A>G p.D359G Chr.19: 582312-582312 on Build GRCh38
1680446 c.536A>G p.D179G Chr.19: 582312-582312 on Build GRCh38
5589844 c.1092C>T p.P364P Chr.19: 582328-582328 on Build GRCh38
3540523 c.530-1G>A p.? Chr.19: 582305-582305 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753155859 Chr.19: 582276-582276 on Build GRCh38
rs576495399 Chr.19: 582329-582329 on Build GRCh38
rs556218809 Chr.19: 582154-582154 on Build GRCh38
rs779976854 Chr.19: 582313-582313 on Build GRCh38
rs146784114 Chr.19: 582314-582314 on Build GRCh38
rs935256511 Chr.19: 582153-582153 on Build GRCh38
rs1054255143 Chr.19: 582254-582254 on Build GRCh38
rs1026754394 Chr.19: 582172-582172 on Build GRCh38
rs536000280 Chr.19: 582145-582145 on Build GRCh38
rs554598015 Chr.19: 582234-582234 on Build GRCh38
rs369856442 Chr.19: 582268-582268 on Build GRCh38
rs781097295 Chr.19: 582296-582296 on Build GRCh38
rs991163862 Chr.19: 582166-582166 on Build GRCh38
rs1035357491 Chr.19: 582160-582160 on Build GRCh38
rs553637912 Chr.19: 582175-582175 on Build GRCh38
rs768559835 Chr.19: 582317-582317 on Build GRCh38
rs757345514 Chr.19: 582293-582293 on Build GRCh38
rs772609315 Chr.19: 582271-582273 on Build GRCh38
rs908768792 Chr.19: 582205-582205 on Build GRCh38
rs923786354 Chr.19: 582299-582299 on Build GRCh38
rs200086920 Chr.19: 582290-582290 on Build GRCh38
rs774143658 Chr.19: 582262-582262 on Build GRCh38
rs944312447 Chr.19: 582233-582233 on Build GRCh38
rs894105830 Chr.19: 582316-582316 on Build GRCh38
rs572647656 Chr.19: 582275-582275 on Build GRCh38
rs774585852 Chr.19: 582297-582299 on Build GRCh38
rs372918193 Chr.19: 582285-582285 on Build GRCh38
rs542552123 Chr.19: 582245-582245 on Build GRCh38
rs761617952 Chr.19: 582263-582263 on Build GRCh38
rs764506893 Chr.19: 582289-582289 on Build GRCh38
rs1027138204 Chr.19: 582324-582324 on Build GRCh38
rs753545807 Chr.19: 582273-582273 on Build GRCh38
rs772819177 Chr.19: 582272-582272 on Build GRCh38
rs773186872 Chr.19: 582227-582227 on Build GRCh38
rs574442476 Chr.19: 582163-582163 on Build GRCh38
rs983000267 Chr.19: 582178-582178 on Build GRCh38
rs1015114957 Chr.19: 582232-582232 on Build GRCh38
rs375929078 Chr.19: 582265-582265 on Build GRCh38
rs4919862 Chr.19: 582253-582253 on Build GRCh38
rs202135216 Chr.19: 582292-582292 on Build GRCh38
rs921232937 Chr.19: 582236-582236 on Build GRCh38
rs770770103 Chr.19: 582308-582310 on Build GRCh38
rs774338462 Chr.19: 582326-582326 on Build GRCh38
rs112152563 Chr.19: 582310-582310 on Build GRCh38
rs1003743321 Chr.19: 582226-582226 on Build GRCh38

View Full Product Details