Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26184481-26184750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAREM2
Entrez Gene ID
150946
Gene Name
GRB2 associated regulator of MAPK1 subtype 2
Gene Aliases
FAM59B, GAREML
UniGene
Hs.187912
Gene Chromosome Location
Chr.2: 26173091-26202405 on Build GRCh38
Transcripts
NM_001168241.1 NM_001191033.1 XM_006711951.3 XM_011532564.2 XM_011532565.2 XM_011532566.2 XM_011532567.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6902793 c.664C>A p.R222S Chr.2: 26184512-26184512 on Build GRCh38
5439148 c.778C>T p.R260C Chr.2: 26184626-26184626 on Build GRCh38
2912127 c.718C>G p.R240G Chr.2: 26184566-26184566 on Build GRCh38
2912125 c.665G>A p.R222H Chr.2: 26184513-26184513 on Build GRCh38
6031937 c.876T>C p.V292V Chr.2: 26184724-26184724 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778477410 Chr.2: 26184682-26184682 on Build GRCh38
rs745349822 Chr.2: 26184695-26184695 on Build GRCh38
rs997360663 Chr.2: 26184595-26184595 on Build GRCh38
rs778045444 Chr.2: 26184581-26184581 on Build GRCh38
rs13011490 Chr.2: 26184650-26184650 on Build GRCh38
rs12992439 Chr.2: 26184621-26184621 on Build GRCh38
rs771439596 Chr.2: 26184731-26184731 on Build GRCh38
rs868073538 Chr.2: 26184626-26184626 on Build GRCh38
rs1004714188 Chr.2: 26184649-26184649 on Build GRCh38
rs936499681 Chr.2: 26184540-26184540 on Build GRCh38
rs563365030 Chr.2: 26184556-26184556 on Build GRCh38
rs749718476 Chr.2: 26184633-26184633 on Build GRCh38
rs974221884 Chr.2: 26184709-26184709 on Build GRCh38
rs900408741 Chr.2: 26184562-26184562 on Build GRCh38
rs963244694 Chr.2: 26184680-26184680 on Build GRCh38
rs1030094532 Chr.2: 26184598-26184598 on Build GRCh38
rs753096677 Chr.2: 26184529-26184529 on Build GRCh38
rs891090595 Chr.2: 26184623-26184623 on Build GRCh38
rs542144215 Chr.2: 26184544-26184544 on Build GRCh38
rs574826604 Chr.2: 26184508-26184508 on Build GRCh38
rs865816282 Chr.2: 26184618-26184618 on Build GRCh38
rs12987199 Chr.2: 26184620-26184620 on Build GRCh38
rs866778702 Chr.2: 26184590-26184590 on Build GRCh38
rs756889040 Chr.2: 26184667-26184667 on Build GRCh38

View Full Product Details