Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 62894934-62895195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHLPP1
Entrez Gene ID
23239
Gene Name
PH domain and leucine rich repeat protein phosphatase 1
Gene Aliases
PHLPP, PLEKHE1, PPM3A, SCOP
UniGene
Hs.465337
Gene Chromosome Location
Chr.18: 62715439-62980443 on Build GRCh38
Transcripts
NM_194449.3 XM_011525886.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7083463 c.2106delT p.L703fs*1 Chr.18: 62895050-62895050 on Build GRCh38
5651179 c.540G>C p.L180F Chr.18: 62895023-62895023 on Build GRCh38
4417818 c.2121A>T p.P707P Chr.18: 62895065-62895065 on Build GRCh38
6221039 c.606C>A p.T202T Chr.18: 62895089-62895089 on Build GRCh38
1324534 c.658G>A p.V220I Chr.18: 62895141-62895141 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745406508 Chr.18: 62895090-62895090 on Build GRCh38
rs753885588 Chr.18: 62895045-62895045 on Build GRCh38
rs943789599 Chr.18: 62895132-62895132 on Build GRCh38
rs769339698 Chr.18: 62895092-62895092 on Build GRCh38
rs757606256 Chr.18: 62895144-62895144 on Build GRCh38
rs891556578 Chr.18: 62895165-62895165 on Build GRCh38
rs751340128 Chr.18: 62895021-62895021 on Build GRCh38
rs758822508 Chr.18: 62895071-62895071 on Build GRCh38
rs746653776 Chr.18: 62894990-62894990 on Build GRCh38
rs760809792 Chr.18: 62895036-62895036 on Build GRCh38
rs777470399 Chr.18: 62894979-62894979 on Build GRCh38
rs759602540 Chr.18: 62895124-62895124 on Build GRCh38
rs779437497 Chr.18: 62895048-62895048 on Build GRCh38
rs755063798 Chr.18: 62895047-62895047 on Build GRCh38
rs376292612 Chr.18: 62895141-62895141 on Build GRCh38
rs775534349 Chr.18: 62895018-62895018 on Build GRCh38
rs779714268 Chr.18: 62895172-62895172 on Build GRCh38
rs755768369 Chr.18: 62895160-62895160 on Build GRCh38
rs776883345 Chr.18: 62895123-62895123 on Build GRCh38
rs761620812 Chr.18: 62895115-62895115 on Build GRCh38
rs762905476 Chr.18: 62895020-62895020 on Build GRCh38
rs569202020 Chr.18: 62895174-62895174 on Build GRCh38
rs747311419 Chr.18: 62895076-62895076 on Build GRCh38
rs749249261 Chr.18: 62895111-62895111 on Build GRCh38
rs371678653 Chr.18: 62895131-62895131 on Build GRCh38
rs766619854 Chr.18: 62894994-62894995 on Build GRCh38
rs772396827 Chr.18: 62894978-62894978 on Build GRCh38
rs745518664 Chr.18: 62895158-62895158 on Build GRCh38
rs778127349 Chr.18: 62895075-62895075 on Build GRCh38
rs377114221 Chr.18: 62895105-62895105 on Build GRCh38
rs776012483 Chr.18: 62895008-62895008 on Build GRCh38
rs780871075 Chr.18: 62895089-62895089 on Build GRCh38
rs759051472 Chr.18: 62895010-62895010 on Build GRCh38
rs774317972 Chr.18: 62895114-62895114 on Build GRCh38
rs770562814 Chr.18: 62895001-62895001 on Build GRCh38
rs573248430 Chr.18: 62894968-62894968 on Build GRCh38
rs919813269 Chr.18: 62894980-62894980 on Build GRCh38
rs766558986 Chr.18: 62895118-62895118 on Build GRCh38
rs780770466 Chr.18: 62895151-62895151 on Build GRCh38
rs778154482 Chr.18: 62895043-62895043 on Build GRCh38
rs369866420 Chr.18: 62895112-62895112 on Build GRCh38
rs770525601 Chr.18: 62895081-62895081 on Build GRCh38
rs758914214 Chr.18: 62895140-62895140 on Build GRCh38
rs201803707 Chr.18: 62895011-62895011 on Build GRCh38
rs624821 Chr.18: 62895065-62895065 on Build GRCh38

View Full Product Details