Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37135123-37135388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL2RB
Entrez Gene ID
3560
Gene Name
interleukin 2 receptor subunit beta
Gene Aliases
CD122, IL15RB, P70-75
UniGene
Hs.474787
Gene Chromosome Location
Chr.22: 37125838-37149922 on Build GRCh38
Transcripts
NM_000878.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3150187 c.812G>T p.G271V Chr.22: 37135334-37135334 on Build GRCh38
5517845 c.810C>T p.T270T Chr.22: 37135336-37135336 on Build GRCh38
1751857 c.802A>G p.R268G Chr.22: 37135344-37135344 on Build GRCh38
4848374 c.795C>A p.I265I Chr.22: 37135351-37135351 on Build GRCh38
6526536 c.771C>T p.F257F Chr.22: 37135375-37135375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369414360 Chr.22: 37135282-37135282 on Build GRCh38
rs149775592 Chr.22: 37135245-37135245 on Build GRCh38
rs1019564610 Chr.22: 37135218-37135221 on Build GRCh38
rs749449499 Chr.22: 37135219-37135220 on Build GRCh38
rs779620990 Chr.22: 37135283-37135283 on Build GRCh38
rs771362307 Chr.22: 37135251-37135260 on Build GRCh38
rs913282117 Chr.22: 37135250-37135251 on Build GRCh38
rs115475423 Chr.22: 37135336-37135336 on Build GRCh38
rs532078306 Chr.22: 37135244-37135244 on Build GRCh38
rs564694416 Chr.22: 37135145-37135145 on Build GRCh38
rs3218306 Chr.22: 37135249-37135249 on Build GRCh38
rs893119583 Chr.22: 37135264-37135264 on Build GRCh38
rs190181994 Chr.22: 37135250-37135250 on Build GRCh38
rs116250343 Chr.22: 37135335-37135335 on Build GRCh38
rs907625144 Chr.22: 37135357-37135357 on Build GRCh38
rs3830742 Chr.22: 37135217-37135218 on Build GRCh38
rs768662546 Chr.22: 37135351-37135351 on Build GRCh38
rs896912294 Chr.22: 37135149-37135149 on Build GRCh38
rs767763441 Chr.22: 37135310-37135312 on Build GRCh38
rs781581480 Chr.22: 37135314-37135314 on Build GRCh38
rs145688666 Chr.22: 37135281-37135281 on Build GRCh38
rs765941615 Chr.22: 37135255-37135255 on Build GRCh38
rs373228367 Chr.22: 37135297-37135297 on Build GRCh38
rs965181639 Chr.22: 37135231-37135231 on Build GRCh38
rs907721456 Chr.22: 37135234-37135234 on Build GRCh38
rs751259673 Chr.22: 37135291-37135291 on Build GRCh38
rs774433459 Chr.22: 37135354-37135354 on Build GRCh38
rs778572562 Chr.22: 37135290-37135290 on Build GRCh38
rs953769732 Chr.22: 37135175-37135175 on Build GRCh38
rs763018682 Chr.22: 37135348-37135348 on Build GRCh38
rs373005180 Chr.22: 37135248-37135249 on Build GRCh38
rs1054439631 Chr.22: 37135260-37135260 on Build GRCh38
rs1010339565 Chr.22: 37135272-37135272 on Build GRCh38
rs995151722 Chr.22: 37135152-37135152 on Build GRCh38
rs771589404 Chr.22: 37135300-37135300 on Build GRCh38
rs777497905 Chr.22: 37135218-37135219 on Build GRCh38
rs1027071019 Chr.22: 37135163-37135163 on Build GRCh38
rs201360122 Chr.22: 37135339-37135339 on Build GRCh38
rs951679721 Chr.22: 37135341-37135341 on Build GRCh38
rs754600437 Chr.22: 37135285-37135285 on Build GRCh38

View Full Product Details