Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 27665889-27666134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR1D
Entrez Gene ID
51082
Gene Name
RNA polymerase I subunit D
Gene Aliases
AC19, POLR1C, RPA16, RPA9, RPAC2, RPC16, RPO1-3, TCS2
UniGene
Hs.507584
Gene Chromosome Location
Chr.13: 27620743-27667422 on Build GRCh38
Transcripts
NM_001206559.1 NM_152705.2 XM_005266414.2 XM_017020622.1 XM_017020623.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5052100 c.369A>T p.*123C Chr.13: 27665953-27665953 on Build GRCh38
6585694 c.362G>C p.R121P Chr.13: 27665946-27665946 on Build GRCh38
5349888 c.362G>A p.R121Q Chr.13: 27665946-27665946 on Build GRCh38
1366178 c.320G>A p.R107Q Chr.13: 27665904-27665904 on Build GRCh38
5739916 c.364C>T p.R122W Chr.13: 27665948-27665948 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772659247 Chr.13: 27666000-27666001 on Build GRCh38
rs773170037 Chr.13: 27665978-27665978 on Build GRCh38
rs776979193 Chr.13: 27665946-27665946 on Build GRCh38
rs141057739 Chr.13: 27665945-27665945 on Build GRCh38
rs767314108 Chr.13: 27665972-27665972 on Build GRCh38
rs557482249 Chr.13: 27666000-27666000 on Build GRCh38
rs910327619 Chr.13: 27665960-27665960 on Build GRCh38
rs75314566 Chr.13: 27665938-27665938 on Build GRCh38
rs892716649 Chr.13: 27666033-27666033 on Build GRCh38
rs1041994326 Chr.13: 27665988-27665988 on Build GRCh38
rs201474841 Chr.13: 27665951-27665951 on Build GRCh38
rs902144870 Chr.13: 27665994-27665994 on Build GRCh38
rs1022703728 Chr.13: 27666103-27666104 on Build GRCh38
rs764415521 Chr.13: 27666014-27666014 on Build GRCh38
rs971936141 Chr.13: 27665924-27665924 on Build GRCh38
rs779185286 Chr.13: 27665966-27665981 on Build GRCh38
rs778356800 Chr.13: 27665930-27665930 on Build GRCh38
rs201427232 Chr.13: 27665948-27665948 on Build GRCh38
rs3742113 Chr.13: 27665922-27665922 on Build GRCh38
rs896247354 Chr.13: 27666007-27666007 on Build GRCh38
rs188886669 Chr.13: 27666003-27666003 on Build GRCh38
rs760445177 Chr.13: 27665984-27665984 on Build GRCh38
rs754522425 Chr.13: 27666000-27666000 on Build GRCh38
rs568728057 Chr.13: 27665990-27665990 on Build GRCh38
rs779645216 Chr.13: 27665912-27665912 on Build GRCh38
rs570345582 Chr.13: 27666100-27666100 on Build GRCh38
rs1034751087 Chr.13: 27665997-27665997 on Build GRCh38
rs758774974 Chr.13: 27665937-27665937 on Build GRCh38
rs757957762 Chr.13: 27666005-27666005 on Build GRCh38
rs1023921095 Chr.13: 27666094-27666094 on Build GRCh38
rs770058121 Chr.13: 27665949-27665949 on Build GRCh38
rs368524929 Chr.13: 27665916-27665916 on Build GRCh38
rs142852632 Chr.13: 27665919-27665919 on Build GRCh38
rs202004435 Chr.13: 27665910-27665910 on Build GRCh38
rs775372461 Chr.13: 27665959-27665959 on Build GRCh38
rs747337734 Chr.13: 27665940-27665940 on Build GRCh38
rs1012066469 Chr.13: 27666050-27666050 on Build GRCh38
rs530426478 Chr.13: 27665995-27665995 on Build GRCh38
rs867786492 Chr.13: 27665941-27665941 on Build GRCh38

View Full Product Details