Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 82663549-82663816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP3B2
Entrez Gene ID
8120
Gene Name
adaptor related protein complex 3 beta 2 subunit
Gene Aliases
NAPTB
UniGene
Hs.199593
Gene Chromosome Location
Chr.15: 82659281-82710112 on Build GRCh38
Transcripts
NM_001278511.1 NM_001278512.1 NM_004644.4 XM_017022640.1 NR_046096.1

Gene Symbol
CPEB1-AS1
Entrez Gene ID
283692
Gene Name
CPEB1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.578667
Gene Chromosome Location
Chr.15: 82647770-82692820 on Build GRCh38
Transcripts
NM_001278511.1 NM_001278512.1 NM_004644.4 XM_017022640.1 NR_046096.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5383925 c.2420C>T p.S807F Chr.15: 82663580-82663580 on Build GRCh38
965706 c.2364C>A p.S788S Chr.15: 82663816-82663816 on Build GRCh38
1183136 c.2425C>T p.L809F Chr.15: 82663575-82663575 on Build GRCh38
4057361 c.2384C>T p.P795L Chr.15: 82663616-82663616 on Build GRCh38
4057360 c.2412G>T p.K804N Chr.15: 82663588-82663588 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750352788 Chr.15: 82663581-82663581 on Build GRCh38
rs759624105 Chr.15: 82663760-82663760 on Build GRCh38
rs879739117 Chr.15: 82663696-82663700 on Build GRCh38
rs773494405 Chr.15: 82663658-82663658 on Build GRCh38
rs954689381 Chr.15: 82663683-82663683 on Build GRCh38
rs772463703 Chr.15: 82663651-82663651 on Build GRCh38
rs970005095 Chr.15: 82663790-82663790 on Build GRCh38
rs756085102 Chr.15: 82663582-82663582 on Build GRCh38
rs559048091 Chr.15: 82663737-82663737 on Build GRCh38
rs201703765 Chr.15: 82663599-82663599 on Build GRCh38
rs371403262 Chr.15: 82663575-82663575 on Build GRCh38
rs1037633603 Chr.15: 82663749-82663749 on Build GRCh38
rs193241037 Chr.15: 82663739-82663739 on Build GRCh38
rs923247894 Chr.15: 82663682-82663682 on Build GRCh38
rs998727894 Chr.15: 82663601-82663601 on Build GRCh38
rs748851002 Chr.15: 82663597-82663597 on Build GRCh38
rs750249098 Chr.15: 82663785-82663785 on Build GRCh38
rs778557732 Chr.15: 82663604-82663604 on Build GRCh38
rs779531557 Chr.15: 82663589-82663589 on Build GRCh38
rs574371408 Chr.15: 82663741-82663741 on Build GRCh38
rs765439454 Chr.15: 82663764-82663764 on Build GRCh38
rs927303042 Chr.15: 82663751-82663751 on Build GRCh38
rs1053342909 Chr.15: 82663664-82663664 on Build GRCh38
rs759594744 Chr.15: 82663669-82663669 on Build GRCh38
rs376059639 Chr.15: 82663776-82663776 on Build GRCh38
rs139183948 Chr.15: 82663695-82663695 on Build GRCh38
rs747781935 Chr.15: 82663605-82663605 on Build GRCh38
rs763132339 Chr.15: 82663770-82663770 on Build GRCh38
rs777291706 Chr.15: 82663755-82663755 on Build GRCh38
rs747550551 Chr.15: 82663663-82663663 on Build GRCh38
rs751739561 Chr.15: 82663747-82663747 on Build GRCh38
rs771624354 Chr.15: 82663665-82663665 on Build GRCh38
rs549740783 Chr.15: 82663686-82663688 on Build GRCh38
rs895918780 Chr.15: 82663700-82663700 on Build GRCh38
rs775731578 Chr.15: 82663765-82663765 on Build GRCh38

View Full Product Details