Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 12206831-12207056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFRSF1B
Entrez Gene ID
7133
Gene Name
TNF receptor superfamily member 1B
Gene Aliases
CD120b, TBPII, TNF-R-II, TNF-R75, TNFBR, TNFR1B, TNFR2, TNFR80, p75, p75TNFR
UniGene
Hs.256278
Gene Chromosome Location
Chr.1: 12166948-12209222 on Build GRCh38
Transcripts
NM_001066.2 XM_011542060.2 XM_011542063.2 XM_017002214.1 XM_017002215.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4406007 c.1270T>G p.S424A Chr.1: 12206904-12206904 on Build GRCh38
6805055 c.1318C>A p.L440M Chr.1: 12206952-12206952 on Build GRCh38
6019437 c.1379C>G p.P460R Chr.1: 12207013-12207013 on Build GRCh38
1687012 c.1215G>A p.M405I Chr.1: 12206849-12206849 on Build GRCh38
5461956 c.1232G>A p.S411N Chr.1: 12206866-12206866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775871058 Chr.1: 12206940-12206940 on Build GRCh38
rs1055074788 Chr.1: 12206875-12206875 on Build GRCh38
rs1002434839 Chr.1: 12206874-12206874 on Build GRCh38
rs200538351 Chr.1: 12206989-12206989 on Build GRCh38
rs754913078 Chr.1: 12206981-12206981 on Build GRCh38
rs776056858 Chr.1: 12206867-12206867 on Build GRCh38
rs753751204 Chr.1: 12206893-12206893 on Build GRCh38
rs766286313 Chr.1: 12206889-12206889 on Build GRCh38
rs761978081 Chr.1: 12206873-12206873 on Build GRCh38
rs770243493 Chr.1: 12206866-12206866 on Build GRCh38
rs139665575 Chr.1: 12206870-12206870 on Build GRCh38
rs774062294 Chr.1: 12206941-12206941 on Build GRCh38
rs759922521 Chr.1: 12206859-12206859 on Build GRCh38
rs371439340 Chr.1: 12206970-12206970 on Build GRCh38
rs149512902 Chr.1: 12206909-12206909 on Build GRCh38
rs373722040 Chr.1: 12206926-12206926 on Build GRCh38
rs955614166 Chr.1: 12207037-12207037 on Build GRCh38
rs773470753 Chr.1: 12206966-12206966 on Build GRCh38
rs773618391 Chr.1: 12206872-12206872 on Build GRCh38
rs774465726 Chr.1: 12206943-12206943 on Build GRCh38
rs762997433 Chr.1: 12206869-12206869 on Build GRCh38
rs531788500 Chr.1: 12206881-12206881 on Build GRCh38
rs748689571 Chr.1: 12206857-12206857 on Build GRCh38
rs371983819 Chr.1: 12207028-12207028 on Build GRCh38
rs767798835 Chr.1: 12206965-12206965 on Build GRCh38
rs375627878 Chr.1: 12207006-12207006 on Build GRCh38
rs777085409 Chr.1: 12206858-12206858 on Build GRCh38
rs746610371 Chr.1: 12207026-12207026 on Build GRCh38
rs200600328 Chr.1: 12206955-12206955 on Build GRCh38
rs747823545 Chr.1: 12206921-12206921 on Build GRCh38
rs368365494 Chr.1: 12207027-12207027 on Build GRCh38
rs902150853 Chr.1: 12206891-12206891 on Build GRCh38
rs770178506 Chr.1: 12206935-12206935 on Build GRCh38
rs752294850 Chr.1: 12206994-12206994 on Build GRCh38
rs1013427348 Chr.1: 12206884-12206884 on Build GRCh38
rs370292777 Chr.1: 12206902-12206902 on Build GRCh38
rs750229037 Chr.1: 12206882-12206882 on Build GRCh38
rs765241635 Chr.1: 12206985-12206985 on Build GRCh38
rs754452683 Chr.1: 12206894-12206894 on Build GRCh38
rs571590743 Chr.1: 12206925-12206925 on Build GRCh38
rs917000076 Chr.1: 12206885-12206885 on Build GRCh38
rs758171297 Chr.1: 12206922-12206922 on Build GRCh38
rs753752076 Chr.1: 12206979-12206979 on Build GRCh38
rs150855008 Chr.1: 12206888-12206888 on Build GRCh38
rs368698604 Chr.1: 12206942-12206942 on Build GRCh38
rs767165937 Chr.1: 12207022-12207022 on Build GRCh38

View Full Product Details