Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 113162919-113163188 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX54
Entrez Gene ID
79039
Gene Name
DEAD-box helicase 54
Gene Aliases
DP97
UniGene
Hs.506861
Gene Chromosome Location
Chr.12: 113157173-113185479 on Build GRCh38
Transcripts
NM_001111322.1 NM_024072.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6641957 c.2077C>T p.R693W Chr.12: 113163136-113163136 on Build GRCh38
7149428 c.2163G>A p.Q721Q Chr.12: 113162964-113162964 on Build GRCh38
291122 c.2130C>T p.G710G Chr.12: 113162997-113162997 on Build GRCh38
3954216 c.2126C>T p.A709V Chr.12: 113163001-113163001 on Build GRCh38
4676875 c.2081+1delG p.? Chr.12: 113163131-113163131 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775551029 Chr.12: 113163045-113163045 on Build GRCh38
rs777863339 Chr.12: 113163131-113163131 on Build GRCh38
rs61740930 Chr.12: 113162996-113162996 on Build GRCh38
rs769860742 Chr.12: 113163032-113163032 on Build GRCh38
rs372345401 Chr.12: 113163109-113163109 on Build GRCh38
rs748376684 Chr.12: 113162943-113162943 on Build GRCh38
rs760323756 Chr.12: 113163049-113163049 on Build GRCh38
rs748285154 Chr.12: 113163030-113163030 on Build GRCh38
rs753458461 Chr.12: 113163059-113163059 on Build GRCh38
rs374730367 Chr.12: 113163136-113163136 on Build GRCh38
rs1013602419 Chr.12: 113163119-113163119 on Build GRCh38
rs745711580 Chr.12: 113163101-113163101 on Build GRCh38
rs761490088 Chr.12: 113163061-113163061 on Build GRCh38
rs377125749 Chr.12: 113163051-113163051 on Build GRCh38
rs61941434 Chr.12: 113163098-113163098 on Build GRCh38
rs201976305 Chr.12: 113162942-113162942 on Build GRCh38
rs146871883 Chr.12: 113163160-113163160 on Build GRCh38
rs770121584 Chr.12: 113163109-113163119 on Build GRCh38
rs780421067 Chr.12: 113163014-113163014 on Build GRCh38
rs755323794 Chr.12: 113163068-113163068 on Build GRCh38
rs749500859 Chr.12: 113163092-113163092 on Build GRCh38
rs753171122 Chr.12: 113163074-113163074 on Build GRCh38
rs753224317 Chr.12: 113163143-113163143 on Build GRCh38
rs761388349 Chr.12: 113163125-113163125 on Build GRCh38
rs767165625 Chr.12: 113162959-113162959 on Build GRCh38
rs751895552 Chr.12: 113163063-113163063 on Build GRCh38
rs985667240 Chr.12: 113162997-113162997 on Build GRCh38
rs768748452 Chr.12: 113162940-113162940 on Build GRCh38
rs762722877 Chr.12: 113162949-113162949 on Build GRCh38
rs772074021 Chr.12: 113163044-113163044 on Build GRCh38
rs368658866 Chr.12: 113163097-113163097 on Build GRCh38
rs553832787 Chr.12: 113163033-113163033 on Build GRCh38
rs774263265 Chr.12: 113163039-113163039 on Build GRCh38
rs370866432 Chr.12: 113162961-113162961 on Build GRCh38
rs747882413 Chr.12: 113162948-113162949 on Build GRCh38
rs756233801 Chr.12: 113163083-113163083 on Build GRCh38
rs10354 Chr.12: 113162992-113162992 on Build GRCh38
rs756572688 Chr.12: 113163146-113163146 on Build GRCh38
rs753501895 Chr.12: 113163003-113163003 on Build GRCh38
rs11564 Chr.12: 113163135-113163135 on Build GRCh38
rs774154228 Chr.12: 113163038-113163038 on Build GRCh38
rs753895243 Chr.12: 113163159-113163159 on Build GRCh38
rs879749875 Chr.12: 113163127-113163127 on Build GRCh38
rs771092287 Chr.12: 113162955-113162955 on Build GRCh38
rs764999719 Chr.12: 113163134-113163134 on Build GRCh38
rs759467235 Chr.12: 113162957-113162957 on Build GRCh38
rs775622263 Chr.12: 113163105-113163105 on Build GRCh38
rs766659106 Chr.12: 113163062-113163062 on Build GRCh38
rs770652501 Chr.12: 113163034-113163034 on Build GRCh38
rs768085133 Chr.12: 113163116-113163116 on Build GRCh38
rs199682901 Chr.12: 113163026-113163026 on Build GRCh38
rs199780486 Chr.12: 113162948-113162948 on Build GRCh38
rs374685656 Chr.12: 113163091-113163091 on Build GRCh38
rs771984152 Chr.12: 113163103-113163103 on Build GRCh38
rs866568580 Chr.12: 113162965-113162965 on Build GRCh38
rs760686446 Chr.12: 113162947-113162947 on Build GRCh38
rs752039864 Chr.12: 113163020-113163020 on Build GRCh38
rs758742181 Chr.12: 113163010-113163010 on Build GRCh38
rs114497647 Chr.12: 113163022-113163022 on Build GRCh38
rs373264271 Chr.12: 113162956-113162956 on Build GRCh38
rs781714079 Chr.12: 113163069-113163069 on Build GRCh38
rs146097905 Chr.12: 113163000-113163000 on Build GRCh38
rs764083182 Chr.12: 113163156-113163156 on Build GRCh38
rs776008708 Chr.12: 113163140-113163140 on Build GRCh38

View Full Product Details