Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 17098349-17098618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MICU3
Entrez Gene ID
286097
Gene Name
mitochondrial calcium uptake family member 3
Gene Aliases
EFHA2
UniGene
Hs.403594
Gene Chromosome Location
Chr.8: 17027238-17133069 on Build GRCh38
Transcripts
NM_181723.2 XM_005273477.2 XM_006716328.2 XM_006716329.2 XM_006716330.2 XM_006716333.3 XM_011544507.2 XM_011544508.2 XM_017013336.1 XM_017013337.1 XM_017013338.1 XM_017013339.1 XM_017013340.1 XM_017013341.1 XM_017013342.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6536472 c.905C>G p.A302G Chr.8: 17098474-17098474 on Build GRCh38
486241 c.902A>G p.D301G Chr.8: 17098471-17098471 on Build GRCh38
6712665 c.946C>T p.R316C Chr.8: 17098515-17098515 on Build GRCh38
145358 c.947G>A p.R316H Chr.8: 17098516-17098516 on Build GRCh38
4934917 c.954A>G p.T318T Chr.8: 17098523-17098523 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017890430 Chr.8: 17098541-17098541 on Build GRCh38
rs750775223 Chr.8: 17098579-17098579 on Build GRCh38
rs771236928 Chr.8: 17098434-17098434 on Build GRCh38
rs762707220 Chr.8: 17098490-17098490 on Build GRCh38
rs746421534 Chr.8: 17098445-17098445 on Build GRCh38
rs377516649 Chr.8: 17098566-17098566 on Build GRCh38
rs772951620 Chr.8: 17098487-17098487 on Build GRCh38
rs772470650 Chr.8: 17098522-17098522 on Build GRCh38
rs774897217 Chr.8: 17098436-17098436 on Build GRCh38
rs1021099482 Chr.8: 17098380-17098385 on Build GRCh38
rs370673348 Chr.8: 17098583-17098583 on Build GRCh38
rs966917708 Chr.8: 17098391-17098391 on Build GRCh38
rs369768353 Chr.8: 17098584-17098584 on Build GRCh38
rs759465150 Chr.8: 17098551-17098551 on Build GRCh38
rs758283207 Chr.8: 17098413-17098413 on Build GRCh38
rs768211230 Chr.8: 17098453-17098453 on Build GRCh38
rs373059368 Chr.8: 17098557-17098557 on Build GRCh38
rs141210355 Chr.8: 17098382-17098382 on Build GRCh38
rs1046742234 Chr.8: 17098386-17098386 on Build GRCh38
rs761401227 Chr.8: 17098458-17098458 on Build GRCh38
rs778110266 Chr.8: 17098513-17098513 on Build GRCh38
rs201094121 Chr.8: 17098419-17098419 on Build GRCh38
rs749698151 Chr.8: 17098429-17098429 on Build GRCh38
rs780632265 Chr.8: 17098531-17098531 on Build GRCh38
rs752721870 Chr.8: 17098558-17098558 on Build GRCh38
rs369698557 Chr.8: 17098548-17098548 on Build GRCh38
rs776012460 Chr.8: 17098454-17098454 on Build GRCh38
rs145026824 Chr.8: 17098403-17098403 on Build GRCh38
rs772831859 Chr.8: 17098545-17098545 on Build GRCh38
rs111567148 Chr.8: 17098508-17098508 on Build GRCh38
rs777268108 Chr.8: 17098590-17098590 on Build GRCh38
rs201128602 Chr.8: 17098517-17098517 on Build GRCh38
rs756364772 Chr.8: 17098509-17098509 on Build GRCh38
rs202243126 Chr.8: 17098515-17098515 on Build GRCh38
rs779213885 Chr.8: 17098516-17098516 on Build GRCh38
rs576616695 Chr.8: 17098585-17098585 on Build GRCh38
rs766373078 Chr.8: 17098418-17098418 on Build GRCh38
rs767381722 Chr.8: 17098556-17098556 on Build GRCh38
rs200222701 Chr.8: 17098506-17098506 on Build GRCh38
rs748312612 Chr.8: 17098423-17098423 on Build GRCh38
rs953883451 Chr.8: 17098442-17098442 on Build GRCh38
rs755065842 Chr.8: 17098421-17098421 on Build GRCh38
rs762654221 Chr.8: 17098547-17098547 on Build GRCh38
rs754110052 Chr.8: 17098574-17098574 on Build GRCh38
rs371590005 Chr.8: 17098425-17098425 on Build GRCh38
rs375641873 Chr.8: 17098499-17098499 on Build GRCh38
rs1044965532 Chr.8: 17098406-17098406 on Build GRCh38
rs779280174 Chr.8: 17098577-17098577 on Build GRCh38
rs892574757 Chr.8: 17098503-17098503 on Build GRCh38
rs766139081 Chr.8: 17098498-17098498 on Build GRCh38
rs1007780083 Chr.8: 17098532-17098532 on Build GRCh38
rs764891589 Chr.8: 17098468-17098468 on Build GRCh38
rs747691787 Chr.8: 17098544-17098544 on Build GRCh38
rs909563797 Chr.8: 17098388-17098388 on Build GRCh38
rs985146618 Chr.8: 17098467-17098467 on Build GRCh38

View Full Product Details