Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 158712105-158712378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EBF1
Entrez Gene ID
1879
Gene Name
early B-cell factor 1
Gene Aliases
COE1, EBF, O/E-1, OLF1
UniGene
Hs.573143
Gene Chromosome Location
Chr.5: 158695915-159099762 on Build GRCh38
Transcripts
NM_001290360.2 NM_001324101.1 NM_001324103.1 NM_001324106.1 NM_001324107.1 NM_001324108.1 NM_001324109.1 NM_001324111.1 NM_024007.4 NM_182708.2 XM_017009192.1 XM_017009193.1 XM_017009194.1 XM_017009195.1 XM_017009196.1 XM_017009197.1 XM_017009198.1 XM_017009199.1 XM_017009200.1 XM_017009201.1 XM_017009204.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3919507 c.1505C>T p.S502F Chr.5: 158712198-158712198 on Build GRCh38
7104203 c.1426C>T p.P476S Chr.5: 158712277-158712277 on Build GRCh38
1435624 c.1509delC p.T504fs*14 Chr.5: 158712194-158712194 on Build GRCh38
6327814 c.1406G>A p.G469E Chr.5: 158712297-158712297 on Build GRCh38
3941222 c.1467C>T p.N489N Chr.5: 158712236-158712236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367971813 Chr.5: 158712299-158712299 on Build GRCh38
rs374533175 Chr.5: 158712168-158712168 on Build GRCh38
rs753073484 Chr.5: 158712203-158712203 on Build GRCh38
rs775144114 Chr.5: 158712264-158712264 on Build GRCh38
rs755222390 Chr.5: 158712140-158712140 on Build GRCh38
rs760492043 Chr.5: 158712181-158712181 on Build GRCh38
rs370909298 Chr.5: 158712142-158712142 on Build GRCh38
rs749298303 Chr.5: 158712351-158712351 on Build GRCh38
rs745516757 Chr.5: 158712169-158712169 on Build GRCh38
rs143146943 Chr.5: 158712293-158712293 on Build GRCh38
rs779578503 Chr.5: 158712347-158712347 on Build GRCh38
rs1000993013 Chr.5: 158712242-158712242 on Build GRCh38
rs769733104 Chr.5: 158712357-158712357 on Build GRCh38
rs181351521 Chr.5: 158712202-158712202 on Build GRCh38
rs776477204 Chr.5: 158712287-158712287 on Build GRCh38
rs751648799 Chr.5: 158712196-158712196 on Build GRCh38
rs781680897 Chr.5: 158712143-158712143 on Build GRCh38
rs748539172 Chr.5: 158712157-158712157 on Build GRCh38
rs267600518 Chr.5: 158712255-158712255 on Build GRCh38
rs147477056 Chr.5: 158712346-158712346 on Build GRCh38
rs913725865 Chr.5: 158712310-158712310 on Build GRCh38
rs756448619 Chr.5: 158712312-158712312 on Build GRCh38
rs745371679 Chr.5: 158712246-158712246 on Build GRCh38
rs768408460 Chr.5: 158712282-158712282 on Build GRCh38
rs372495632 Chr.5: 158712315-158712315 on Build GRCh38
rs771633394 Chr.5: 158712175-158712175 on Build GRCh38
rs755170406 Chr.5: 158712200-158712200 on Build GRCh38
rs757758762 Chr.5: 158712245-158712245 on Build GRCh38
rs776525933 Chr.5: 158712193-158712193 on Build GRCh38
rs754162618 Chr.5: 158712329-158712329 on Build GRCh38
rs748700592 Chr.5: 158712162-158712162 on Build GRCh38
rs554904637 Chr.5: 158712218-158712218 on Build GRCh38
rs770456688 Chr.5: 158712166-158712166 on Build GRCh38
rs780983339 Chr.5: 158712354-158712354 on Build GRCh38
rs747900765 Chr.5: 158712355-158712355 on Build GRCh38
rs367836384 Chr.5: 158712194-158712194 on Build GRCh38
rs770360981 Chr.5: 158712161-158712161 on Build GRCh38
rs115365806 Chr.5: 158712131-158712131 on Build GRCh38
rs374891820 Chr.5: 158712253-158712253 on Build GRCh38
rs574867896 Chr.5: 158712311-158712311 on Build GRCh38
rs763968946 Chr.5: 158712182-158712182 on Build GRCh38
rs768355410 Chr.5: 158712353-158712353 on Build GRCh38
rs778335068 Chr.5: 158712221-158712221 on Build GRCh38
rs556574598 Chr.5: 158712306-158712306 on Build GRCh38
rs146847386 Chr.5: 158712220-158712220 on Build GRCh38
rs779316113 Chr.5: 158712345-158712345 on Build GRCh38
rs761796846 Chr.5: 158712288-158712288 on Build GRCh38
rs749775088 Chr.5: 158712227-158712227 on Build GRCh38
rs757712452 Chr.5: 158712338-158712338 on Build GRCh38
rs771271567 Chr.5: 158712311-158712313 on Build GRCh38
rs775195432 Chr.5: 158712176-158712176 on Build GRCh38
rs746711236 Chr.5: 158712277-158712277 on Build GRCh38

View Full Product Details