Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 104994563-104994790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf45
Entrez Gene ID
121053
Gene Name
chromosome 12 open reading frame 45
Gene Aliases
-
UniGene
Hs.295563
Gene Chromosome Location
Chr.12: 104986320-104994727 on Build GRCh38
Transcripts
NM_152318.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
297980 c.544delA p.K184fs*>2 Chr.12: 104994682-104994682 on Build GRCh38
7303721 c.522A>T p.E174D Chr.12: 104994660-104994660 on Build GRCh38
6614198 c.484A>G p.K162E Chr.12: 104994622-104994622 on Build GRCh38
3455397 c.471C>T p.T157T Chr.12: 104994609-104994609 on Build GRCh38
6071137 c.486G>T p.K162N Chr.12: 104994624-104994624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760572551 Chr.12: 104994680-104994690 on Build GRCh38
rs745896625 Chr.12: 104994590-104994590 on Build GRCh38
rs747222960 Chr.12: 104994611-104994611 on Build GRCh38
rs202087919 Chr.12: 104994661-104994661 on Build GRCh38
rs1033149521 Chr.12: 104994757-104994757 on Build GRCh38
rs777032057 Chr.12: 104994681-104994682 on Build GRCh38
rs538963565 Chr.12: 104994698-104994698 on Build GRCh38
rs759124890 Chr.12: 104994688-104994688 on Build GRCh38
rs767316407 Chr.12: 104994692-104994692 on Build GRCh38
rs548987873 Chr.12: 104994758-104994758 on Build GRCh38
rs184257841 Chr.12: 104994670-104994670 on Build GRCh38
rs758473392 Chr.12: 104994608-104994608 on Build GRCh38
rs377159683 Chr.12: 104994682-104994682 on Build GRCh38
rs369460633 Chr.12: 104994632-104994632 on Build GRCh38
rs75497332 Chr.12: 104994767-104994767 on Build GRCh38
rs369173579 Chr.12: 104994694-104994694 on Build GRCh38
rs770159752 Chr.12: 104994645-104994645 on Build GRCh38
rs781713189 Chr.12: 104994729-104994729 on Build GRCh38
rs1056870624 Chr.12: 104994630-104994630 on Build GRCh38
rs780475223 Chr.12: 104994610-104994610 on Build GRCh38
rs996031455 Chr.12: 104994757-104994758 on Build GRCh38
rs749344966 Chr.12: 104994733-104994733 on Build GRCh38
rs373932826 Chr.12: 104994707-104994707 on Build GRCh38
rs769146367 Chr.12: 104994614-104994614 on Build GRCh38
rs777075411 Chr.12: 104994618-104994618 on Build GRCh38
rs78239531 Chr.12: 104994721-104994721 on Build GRCh38
rs751843461 Chr.12: 104994710-104994710 on Build GRCh38
rs765859693 Chr.12: 104994680-104994680 on Build GRCh38
rs756111844 Chr.12: 104994696-104994696 on Build GRCh38
rs376760830 Chr.12: 104994595-104994595 on Build GRCh38
rs141562989 Chr.12: 104994706-104994706 on Build GRCh38
rs763974133 Chr.12: 104994680-104994681 on Build GRCh38
rs748498088 Chr.12: 104994730-104994730 on Build GRCh38
rs920509447 Chr.12: 104994591-104994591 on Build GRCh38

View Full Product Details