Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129001991-129002218 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCC1
Entrez Gene ID
79825
Gene Name
EF-hand and coiled-coil domain containing 1
Gene Aliases
C3orf73, CCDC48
UniGene
Hs.134807
Gene Chromosome Location
Chr.3: 129001604-129040742 on Build GRCh38
Transcripts
NM_024768.2 XM_005247781.3 XM_011513163.2 XM_011513050.2 XM_017006939.1 XM_017006940.1 XM_017006941.1 XM_017006942.1 XM_017006943.1 XM_017006945.1 XM_017006946.1 XM_017006947.1 XM_017006952.1

Gene Symbol
KIAA1257
Entrez Gene ID
57501
Gene Name
KIAA1257
Gene Aliases
-
UniGene
Hs.518247
Gene Chromosome Location
Chr.3: 128910186-129032431 on Build GRCh38
Transcripts
NM_024768.2 XM_005247781.3 XM_011513163.2 XM_011513050.2 XM_017006939.1 XM_017006940.1 XM_017006941.1 XM_017006942.1 XM_017006943.1 XM_017006945.1 XM_017006946.1 XM_017006947.1 XM_017006952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4987902 c.400C>T p.R134C Chr.3: 129002028-129002028 on Build GRCh38
4952097 c.409C>A p.P137T Chr.3: 129002037-129002037 on Build GRCh38
6390980 c.418C>T p.L140F Chr.3: 129002046-129002046 on Build GRCh38
6564231 c.498C>T p.L166L Chr.3: 129002126-129002126 on Build GRCh38
6431893 c.586G>A p.D196N Chr.3: 129002214-129002214 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886067135 Chr.3: 129002044-129002044 on Build GRCh38
rs979811606 Chr.3: 129002041-129002041 on Build GRCh38
rs1022674399 Chr.3: 129002161-129002161 on Build GRCh38
rs889340325 Chr.3: 129002136-129002136 on Build GRCh38
rs767548537 Chr.3: 129002113-129002113 on Build GRCh38
rs554623904 Chr.3: 129002051-129002051 on Build GRCh38
rs541157948 Chr.3: 129002068-129002068 on Build GRCh38
rs563646191 Chr.3: 129002164-129002164 on Build GRCh38
rs868335940 Chr.3: 129002184-129002184 on Build GRCh38
rs959855891 Chr.3: 129002015-129002015 on Build GRCh38
rs768647320 Chr.3: 129002075-129002075 on Build GRCh38
rs923391278 Chr.3: 129002066-129002066 on Build GRCh38
rs933468932 Chr.3: 129002111-129002111 on Build GRCh38
rs760546571 Chr.3: 129002154-129002154 on Build GRCh38
rs542109937 Chr.3: 129002143-129002143 on Build GRCh38
rs531145864 Chr.3: 129002167-129002167 on Build GRCh38
rs780135100 Chr.3: 129002075-129002075 on Build GRCh38
rs746289975 Chr.3: 129002063-129002063 on Build GRCh38
rs778297815 Chr.3: 129002031-129002031 on Build GRCh38
rs1018621130 Chr.3: 129002080-129002080 on Build GRCh38
rs552349632 Chr.3: 129002188-129002188 on Build GRCh38
rs769247574 Chr.3: 129002165-129002166 on Build GRCh38
rs1023433722 Chr.3: 129002187-129002195 on Build GRCh38
rs753569809 Chr.3: 129002175-129002175 on Build GRCh38
rs182074815 Chr.3: 129002030-129002030 on Build GRCh38
rs559054712 Chr.3: 129002130-129002130 on Build GRCh38
rs780841462 Chr.3: 129002064-129002064 on Build GRCh38
rs763036432 Chr.3: 129002166-129002166 on Build GRCh38
rs574355302 Chr.3: 129002093-129002093 on Build GRCh38
rs747038490 Chr.3: 129002166-129002168 on Build GRCh38
rs761633622 Chr.3: 129002112-129002112 on Build GRCh38
rs762965217 Chr.3: 129002072-129002072 on Build GRCh38
rs1050646500 Chr.3: 129002116-129002116 on Build GRCh38
rs993155540 Chr.3: 129002182-129002182 on Build GRCh38
rs529712866 Chr.3: 129002139-129002139 on Build GRCh38
rs747504557 Chr.3: 129002043-129002043 on Build GRCh38
rs771513415 Chr.3: 129002048-129002048 on Build GRCh38
rs991376743 Chr.3: 129002028-129002028 on Build GRCh38
rs925276827 Chr.3: 129002045-129002045 on Build GRCh38
rs996007312 Chr.3: 129002138-129002138 on Build GRCh38

View Full Product Details