Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 8440891-8441126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMT44
Entrez Gene ID
152992
Gene Name
tRNA methyltransferase 44 homolog (S. cerevisiae)
Gene Aliases
C4orf23, METTL19, TRM44
UniGene
Hs.566191
Gene Chromosome Location
Chr.4: 8440774-8512531 on Build GRCh38
Transcripts
NM_152544.2 XM_005247950.3 XM_011513406.2 XM_011513407.2 XM_011513408.1 XM_011513409.2 XM_011513410.2 XM_017007811.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5697955 c.120C>A p.G40G Chr.4: 8440942-8440942 on Build GRCh38
6435716 c.148G>A p.A50T Chr.4: 8440970-8440970 on Build GRCh38
5818206 c.166G>T p.A56S Chr.4: 8440988-8440988 on Build GRCh38
1619161 c.132G>C p.E44D Chr.4: 8440954-8440954 on Build GRCh38
7003758 c.226C>A p.Q76K Chr.4: 8441048-8441048 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771835830 Chr.4: 8441072-8441072 on Build GRCh38
rs748917670 Chr.4: 8441039-8441039 on Build GRCh38
rs894171272 Chr.4: 8440913-8440913 on Build GRCh38
rs758774873 Chr.4: 8440938-8440938 on Build GRCh38
rs773190135 Chr.4: 8441080-8441080 on Build GRCh38
rs974469579 Chr.4: 8441002-8441002 on Build GRCh38
rs955495737 Chr.4: 8440939-8440939 on Build GRCh38
rs547899288 Chr.4: 8440947-8440947 on Build GRCh38
rs771312952 Chr.4: 8441016-8441016 on Build GRCh38
rs137937521 Chr.4: 8440936-8440936 on Build GRCh38
rs184964039 Chr.4: 8441061-8441061 on Build GRCh38
rs768450088 Chr.4: 8441048-8441048 on Build GRCh38
rs778350913 Chr.4: 8440956-8440956 on Build GRCh38
rs964078370 Chr.4: 8440984-8440984 on Build GRCh38
rs564114043 Chr.4: 8441055-8441055 on Build GRCh38
rs1018518544 Chr.4: 8440948-8440948 on Build GRCh38
rs533050698 Chr.4: 8440966-8440966 on Build GRCh38
rs760581652 Chr.4: 8441091-8441091 on Build GRCh38
rs928501910 Chr.4: 8441068-8441068 on Build GRCh38
rs540073637 Chr.4: 8441095-8441095 on Build GRCh38
rs1011657696 Chr.4: 8440925-8440925 on Build GRCh38
rs939131288 Chr.4: 8441099-8441099 on Build GRCh38
rs778924433 Chr.4: 8440928-8440928 on Build GRCh38
rs1008512865 Chr.4: 8440940-8440940 on Build GRCh38
rs560407049 Chr.4: 8440963-8440963 on Build GRCh38
rs974714975 Chr.4: 8440974-8440974 on Build GRCh38
rs927738808 Chr.4: 8441093-8441093 on Build GRCh38
rs919901636 Chr.4: 8441012-8441012 on Build GRCh38
rs758445075 Chr.4: 8440929-8440929 on Build GRCh38
rs752952184 Chr.4: 8440915-8440915 on Build GRCh38
rs762808462 Chr.4: 8441083-8441083 on Build GRCh38
rs781458066 Chr.4: 8441023-8441023 on Build GRCh38
rs989382425 Chr.4: 8441059-8441059 on Build GRCh38
rs527445129 Chr.4: 8441001-8441001 on Build GRCh38
rs750302647 Chr.4: 8441017-8441017 on Build GRCh38
rs902940185 Chr.4: 8440934-8440934 on Build GRCh38
rs766169556 Chr.4: 8441101-8441101 on Build GRCh38

View Full Product Details