Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 17104328-17104583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MICU3
Entrez Gene ID
286097
Gene Name
mitochondrial calcium uptake family member 3
Gene Aliases
EFHA2
UniGene
Hs.403594
Gene Chromosome Location
Chr.8: 17027238-17133069 on Build GRCh38
Transcripts
NM_181723.2 XM_005273477.2 XM_006716328.2 XM_006716329.2 XM_006716330.2 XM_011544507.2 XM_017013336.1 XM_017013337.1 XM_017013340.1 XM_017013342.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2872976 c.985C>T p.R329C Chr.8: 17104391-17104391 on Build GRCh38
3924938 c.1072G>A p.E358K Chr.8: 17104478-17104478 on Build GRCh38
4150686 c.997A>G p.I333V Chr.8: 17104403-17104403 on Build GRCh38
4568150 c.1007T>C p.L336S Chr.8: 17104413-17104413 on Build GRCh38
6322923 c.1015G>T p.D339Y Chr.8: 17104421-17104421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982716276 Chr.8: 17104422-17104422 on Build GRCh38
rs953765889 Chr.8: 17104464-17104464 on Build GRCh38
rs761064458 Chr.8: 17104379-17104379 on Build GRCh38
rs763110983 Chr.8: 17104519-17104519 on Build GRCh38
rs773681873 Chr.8: 17104378-17104378 on Build GRCh38
rs753295478 Chr.8: 17104468-17104468 on Build GRCh38
rs768612883 Chr.8: 17104491-17104491 on Build GRCh38
rs776733635 Chr.8: 17104415-17104415 on Build GRCh38
rs201776772 Chr.8: 17104391-17104391 on Build GRCh38
rs752059291 Chr.8: 17104379-17104379 on Build GRCh38
rs866300143 Chr.8: 17104439-17104439 on Build GRCh38
rs555688593 Chr.8: 17104384-17104384 on Build GRCh38
rs374645079 Chr.8: 17104392-17104392 on Build GRCh38
rs763304294 Chr.8: 17104429-17104429 on Build GRCh38
rs746832652 Chr.8: 17104488-17104488 on Build GRCh38
rs752779729 Chr.8: 17104506-17104506 on Build GRCh38
rs766791079 Chr.8: 17104376-17104376 on Build GRCh38
rs762046314 Chr.8: 17104367-17104367 on Build GRCh38
rs573324672 Chr.8: 17104378-17104379 on Build GRCh38
rs768737632 Chr.8: 17104414-17104414 on Build GRCh38
rs769762241 Chr.8: 17104503-17104503 on Build GRCh38
rs775432229 Chr.8: 17104405-17104405 on Build GRCh38
rs748178195 Chr.8: 17104499-17104499 on Build GRCh38
rs939379138 Chr.8: 17104512-17104512 on Build GRCh38
rs965201399 Chr.8: 17104524-17104524 on Build GRCh38
rs765410229 Chr.8: 17104425-17104425 on Build GRCh38
rs150676145 Chr.8: 17104528-17104528 on Build GRCh38
rs375409276 Chr.8: 17104540-17104540 on Build GRCh38
rs913897577 Chr.8: 17104521-17104521 on Build GRCh38
rs777213150 Chr.8: 17104383-17104383 on Build GRCh38
rs765409458 Chr.8: 17104370-17104370 on Build GRCh38
rs759688512 Chr.8: 17104454-17104455 on Build GRCh38
rs146521840 Chr.8: 17104403-17104403 on Build GRCh38
rs759387770 Chr.8: 17104549-17104549 on Build GRCh38
rs765494279 Chr.8: 17104501-17104501 on Build GRCh38
rs188942953 Chr.8: 17104544-17104544 on Build GRCh38
rs751940168 Chr.8: 17104454-17104454 on Build GRCh38
rs755434329 Chr.8: 17104461-17104461 on Build GRCh38
rs756828221 Chr.8: 17104387-17104387 on Build GRCh38
rs750701200 Chr.8: 17104375-17104375 on Build GRCh38
rs778551274 Chr.8: 17104469-17104469 on Build GRCh38
rs776414512 Chr.8: 17104493-17104493 on Build GRCh38
rs1017738627 Chr.8: 17104390-17104390 on Build GRCh38
rs112454980 Chr.8: 17104409-17104409 on Build GRCh38
rs141934394 Chr.8: 17104514-17104514 on Build GRCh38
rs199588638 Chr.8: 17104452-17104452 on Build GRCh38
rs761772626 Chr.8: 17104417-17104417 on Build GRCh38
rs745443351 Chr.8: 17104475-17104475 on Build GRCh38
rs909813617 Chr.8: 17104436-17104436 on Build GRCh38
rs184496119 Chr.8: 17104530-17104530 on Build GRCh38
rs763066305 Chr.8: 17104509-17104510 on Build GRCh38
rs951154615 Chr.8: 17104418-17104418 on Build GRCh38
rs758140362 Chr.8: 17104480-17104480 on Build GRCh38
rs746886073 Chr.8: 17104413-17104413 on Build GRCh38
rs35902397 Chr.8: 17104447-17104447 on Build GRCh38
rs766670510 Chr.8: 17104444-17104444 on Build GRCh38
rs779947528 Chr.8: 17104487-17104487 on Build GRCh38
rs377387747 Chr.8: 17104363-17104363 on Build GRCh38
rs776931218 Chr.8: 17104449-17104452 on Build GRCh38
rs755487319 Chr.8: 17104382-17104382 on Build GRCh38

View Full Product Details