Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 32595843-32596104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3M
Entrez Gene ID
10480
Gene Name
eukaryotic translation initiation factor 3 subunit M
Gene Aliases
B5, GA17, PCID1, TANGO7, hfl-B5
UniGene
Hs.502244
Gene Chromosome Location
Chr.11: 32583767-32602873 on Build GRCh38
Transcripts
NM_001307929.1 NM_006360.5 XM_011519840.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6652892 c.721T>G p.L241V Chr.11: 32595969-32595969 on Build GRCh38
4770612 c.790G>A p.D264N Chr.11: 32596038-32596038 on Build GRCh38
4191100 c.738T>C p.S246S Chr.11: 32595986-32595986 on Build GRCh38
6652893 c.718C>A p.L240I Chr.11: 32595966-32595966 on Build GRCh38
926372 c.748G>A p.A250T Chr.11: 32595996-32595996 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529945180 Chr.11: 32595929-32595929 on Build GRCh38
rs147860021 Chr.11: 32595878-32595882 on Build GRCh38
rs754977095 Chr.11: 32595974-32595974 on Build GRCh38
rs771733365 Chr.11: 32596055-32596055 on Build GRCh38
rs1019670453 Chr.11: 32595923-32595923 on Build GRCh38
rs757159807 Chr.11: 32596009-32596009 on Build GRCh38
rs147156038 Chr.11: 32596004-32596004 on Build GRCh38
rs758728945 Chr.11: 32595957-32595957 on Build GRCh38
rs747713403 Chr.11: 32595943-32595944 on Build GRCh38
rs778244154 Chr.11: 32595942-32595943 on Build GRCh38
rs751735055 Chr.11: 32595972-32595972 on Build GRCh38
rs781692502 Chr.11: 32596050-32596051 on Build GRCh38
rs143517336 Chr.11: 32595998-32595998 on Build GRCh38
rs368024138 Chr.11: 32596075-32596075 on Build GRCh38
rs146792978 Chr.11: 32595892-32595892 on Build GRCh38
rs771404131 Chr.11: 32596048-32596049 on Build GRCh38
rs1048013525 Chr.11: 32596020-32596020 on Build GRCh38
rs766663936 Chr.11: 32595958-32595958 on Build GRCh38
rs745622122 Chr.11: 32596040-32596040 on Build GRCh38
rs139574814 Chr.11: 32595894-32595894 on Build GRCh38
rs1004237092 Chr.11: 32595900-32595900 on Build GRCh38
rs1037319432 Chr.11: 32595917-32595917 on Build GRCh38
rs140407912 Chr.11: 32596015-32596015 on Build GRCh38
rs746596193 Chr.11: 32596077-32596077 on Build GRCh38
rs778681512 Chr.11: 32596018-32596018 on Build GRCh38

View Full Product Details