Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37137502-37137775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL2RB
Entrez Gene ID
3560
Gene Name
interleukin 2 receptor subunit beta
Gene Aliases
CD122, IL15RB, P70-75
UniGene
Hs.474787
Gene Chromosome Location
Chr.22: 37125838-37149922 on Build GRCh38
Transcripts
NM_000878.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4103913 c.447C>T p.N149N Chr.22: 37137677-37137677 on Build GRCh38
5393573 c.529_530delAC p.T177fs*28 Chr.22: 37137594-37137595 on Build GRCh38
135919 c.455G>A p.W152* Chr.22: 37137669-37137669 on Build GRCh38
6438561 c.509_510GG>AA p.R170Q Chr.22: 37137614-37137615 on Build GRCh38
240225 c.392G>A p.R131H Chr.22: 37137732-37137732 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138490007 Chr.22: 37137611-37137611 on Build GRCh38
rs3218291 Chr.22: 37137524-37137524 on Build GRCh38
rs781374953 Chr.22: 37137733-37137733 on Build GRCh38
rs763844954 Chr.22: 37137546-37137546 on Build GRCh38
rs200697945 Chr.22: 37137743-37137743 on Build GRCh38
rs753138097 Chr.22: 37137576-37137576 on Build GRCh38
rs200381499 Chr.22: 37137625-37137625 on Build GRCh38
rs371794346 Chr.22: 37137653-37137653 on Build GRCh38
rs765775857 Chr.22: 37137678-37137678 on Build GRCh38
rs765538788 Chr.22: 37137570-37137570 on Build GRCh38
rs774119548 Chr.22: 37137619-37137619 on Build GRCh38
rs368501031 Chr.22: 37137606-37137606 on Build GRCh38
rs228957 Chr.22: 37137746-37137746 on Build GRCh38
rs199729784 Chr.22: 37137578-37137578 on Build GRCh38
rs746806656 Chr.22: 37137730-37137730 on Build GRCh38
rs760228358 Chr.22: 37137756-37137756 on Build GRCh38
rs867355828 Chr.22: 37137713-37137713 on Build GRCh38
rs761376327 Chr.22: 37137622-37137622 on Build GRCh38
rs370822006 Chr.22: 37137548-37137548 on Build GRCh38
rs752982148 Chr.22: 37137697-37137697 on Build GRCh38
rs904059541 Chr.22: 37137623-37137623 on Build GRCh38
rs367622513 Chr.22: 37137749-37137749 on Build GRCh38
rs763434888 Chr.22: 37137710-37137710 on Build GRCh38
rs745746134 Chr.22: 37137608-37137608 on Build GRCh38
rs768430184 Chr.22: 37137750-37137750 on Build GRCh38
rs372164885 Chr.22: 37137752-37137752 on Build GRCh38
rs149258462 Chr.22: 37137665-37137665 on Build GRCh38
rs961230216 Chr.22: 37137738-37137738 on Build GRCh38
rs757429668 Chr.22: 37137590-37137590 on Build GRCh38
rs768177172 Chr.22: 37137616-37137616 on Build GRCh38
rs368499991 Chr.22: 37137541-37137541 on Build GRCh38
rs228958 Chr.22: 37137755-37137755 on Build GRCh38
rs749152355 Chr.22: 37137615-37137615 on Build GRCh38
rs764222346 Chr.22: 37137718-37137718 on Build GRCh38
rs771510617 Chr.22: 37137651-37137651 on Build GRCh38
rs781429683 Chr.22: 37137604-37137604 on Build GRCh38
rs766866943 Chr.22: 37137566-37137566 on Build GRCh38
rs773707305 Chr.22: 37137547-37137547 on Build GRCh38
rs200356597 Chr.22: 37137567-37137567 on Build GRCh38
rs750566804 Chr.22: 37137612-37137612 on Build GRCh38
rs751581549 Chr.22: 37137589-37137589 on Build GRCh38
rs758517823 Chr.22: 37137577-37137577 on Build GRCh38
rs201441730 Chr.22: 37137723-37137723 on Build GRCh38

View Full Product Details