Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 50596047-50596320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM65C
Entrez Gene ID
140876
Gene Name
family with sequence similarity 65 member C
Gene Aliases
C20orf175, C20orf176
UniGene
Hs.372578
Gene Chromosome Location
Chr.20: 50586108-50692004 on Build GRCh38
Transcripts
NM_001290268.1 NM_080829.3 NR_110890.1 XM_005260294.3 XM_006723713.3 XM_011528578.2 XM_011528579.2 XM_011528580.2 XM_011528581.2 XM_011528584.2 XM_011528585.2 XM_011528586.2 XM_017027681.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6236149 c.1786C>T p.R596W Chr.20: 50596256-50596256 on Build GRCh38
5443050 c.1818G>C p.A606A Chr.20: 50596224-50596224 on Build GRCh38
5040097 c.1790C>T p.P597L Chr.20: 50596252-50596252 on Build GRCh38
1307516 c.1866G>A p.L622L Chr.20: 50596176-50596176 on Build GRCh38
6660035 c.1871T>C p.V624A Chr.20: 50596171-50596171 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377276576 Chr.20: 50596243-50596243 on Build GRCh38
rs970324353 Chr.20: 50596102-50596102 on Build GRCh38
rs770229488 Chr.20: 50596250-50596250 on Build GRCh38
rs747296395 Chr.20: 50596269-50596273 on Build GRCh38
rs759748109 Chr.20: 50596271-50596271 on Build GRCh38
rs993935175 Chr.20: 50596258-50596258 on Build GRCh38
rs771258806 Chr.20: 50596281-50596282 on Build GRCh38
rs778280983 Chr.20: 50596119-50596119 on Build GRCh38
rs577491042 Chr.20: 50596107-50596107 on Build GRCh38
rs372472260 Chr.20: 50596224-50596224 on Build GRCh38
rs556905789 Chr.20: 50596090-50596090 on Build GRCh38
rs773729956 Chr.20: 50596251-50596251 on Build GRCh38
rs755953536 Chr.20: 50596203-50596203 on Build GRCh38
rs189232949 Chr.20: 50596225-50596225 on Build GRCh38
rs771635715 Chr.20: 50596139-50596139 on Build GRCh38
rs150387614 Chr.20: 50596067-50596067 on Build GRCh38
rs891561427 Chr.20: 50596081-50596081 on Build GRCh38
rs766540580 Chr.20: 50596190-50596190 on Build GRCh38
rs1036057713 Chr.20: 50596169-50596169 on Build GRCh38
rs373141664 Chr.20: 50596240-50596240 on Build GRCh38
rs747842460 Chr.20: 50596225-50596226 on Build GRCh38
rs777729833 Chr.20: 50596105-50596105 on Build GRCh38
rs1042648261 Chr.20: 50596075-50596075 on Build GRCh38
rs761546880 Chr.20: 50596297-50596297 on Build GRCh38
rs769107803 Chr.20: 50596158-50596158 on Build GRCh38
rs373812762 Chr.20: 50596196-50596196 on Build GRCh38
rs373756359 Chr.20: 50596101-50596102 on Build GRCh38
rs76945848 Chr.20: 50596210-50596210 on Build GRCh38
rs765713430 Chr.20: 50596184-50596184 on Build GRCh38
rs747405056 Chr.20: 50596151-50596151 on Build GRCh38
rs372985228 Chr.20: 50596239-50596239 on Build GRCh38
rs757881867 Chr.20: 50596236-50596236 on Build GRCh38
rs1005433558 Chr.20: 50596290-50596290 on Build GRCh38
rs544572870 Chr.20: 50596109-50596109 on Build GRCh38
rs1052679026 Chr.20: 50596079-50596079 on Build GRCh38
rs888325507 Chr.20: 50596282-50596282 on Build GRCh38
rs948126174 Chr.20: 50596188-50596188 on Build GRCh38
rs997176187 Chr.20: 50596073-50596073 on Build GRCh38
rs748469823 Chr.20: 50596249-50596249 on Build GRCh38
rs771568258 Chr.20: 50596240-50596242 on Build GRCh38
rs752458733 Chr.20: 50596091-50596091 on Build GRCh38
rs116264080 Chr.20: 50596261-50596261 on Build GRCh38
rs564876086 Chr.20: 50596202-50596202 on Build GRCh38
rs117579288 Chr.20: 50596098-50596098 on Build GRCh38
rs369019040 Chr.20: 50596208-50596208 on Build GRCh38
rs765113769 Chr.20: 50596301-50596301 on Build GRCh38
rs376015124 Chr.20: 50596293-50596293 on Build GRCh38
rs756817507 Chr.20: 50596117-50596117 on Build GRCh38
rs766461315 Chr.20: 50596255-50596255 on Build GRCh38
rs763041379 Chr.20: 50596185-50596185 on Build GRCh38
rs773648496 Chr.20: 50596242-50596243 on Build GRCh38
rs6126047 Chr.20: 50596140-50596140 on Build GRCh38
rs767937393 Chr.20: 50596069-50596069 on Build GRCh38
rs767658021 Chr.20: 50596284-50596284 on Build GRCh38
rs762407181 Chr.20: 50596171-50596171 on Build GRCh38
rs376080684 Chr.20: 50596226-50596226 on Build GRCh38
rs745318441 Chr.20: 50596127-50596127 on Build GRCh38
rs369926463 Chr.20: 50596166-50596166 on Build GRCh38

View Full Product Details