Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33206300-33206557 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B8
Entrez Gene ID
7923
Gene Name
hydroxysteroid 17-beta dehydrogenase 8
Gene Aliases
D6S2245E, FABG, FABGL, H2-KE6, HKE6, KE6, RING2, SDR30C1, dJ1033B10.9
UniGene
Hs.415058
Gene Chromosome Location
Chr.6: 33204637-33206831 on Build GRCh38
Transcripts
NM_014234.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
219632 c.769+5G>A p.? Chr.6: 33206454-33206454 on Build GRCh38
4776700 c.727G>A p.E243K Chr.6: 33206407-33206407 on Build GRCh38
3702983 c.769+10C>T p.? Chr.6: 33206459-33206459 on Build GRCh38
1621509 c.745A>G p.T249A Chr.6: 33206425-33206425 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753635683 Chr.6: 33206356-33206356 on Build GRCh38
rs775760809 Chr.6: 33206422-33206422 on Build GRCh38
rs149130105 Chr.6: 33206450-33206450 on Build GRCh38
rs955803907 Chr.6: 33206513-33206513 on Build GRCh38
rs747153610 Chr.6: 33206412-33206412 on Build GRCh38
rs749634614 Chr.6: 33206438-33206438 on Build GRCh38
rs779266895 Chr.6: 33206402-33206420 on Build GRCh38
rs955863145 Chr.6: 33206323-33206323 on Build GRCh38
rs767157312 Chr.6: 33206326-33206326 on Build GRCh38
rs749983905 Chr.6: 33206332-33206332 on Build GRCh38
rs573450477 Chr.6: 33206501-33206501 on Build GRCh38
rs778979432 Chr.6: 33206400-33206400 on Build GRCh38
rs755014412 Chr.6: 33206392-33206392 on Build GRCh38
rs770229626 Chr.6: 33206421-33206421 on Build GRCh38
rs772946917 Chr.6: 33206498-33206498 on Build GRCh38
rs201422899 Chr.6: 33206470-33206470 on Build GRCh38
rs375153877 Chr.6: 33206525-33206525 on Build GRCh38
rs367921166 Chr.6: 33206486-33206486 on Build GRCh38
rs761323980 Chr.6: 33206477-33206477 on Build GRCh38
rs746030709 Chr.6: 33206418-33206418 on Build GRCh38
rs748748639 Chr.6: 33206462-33206463 on Build GRCh38
rs540925338 Chr.6: 33206325-33206325 on Build GRCh38
rs1027171631 Chr.6: 33206339-33206339 on Build GRCh38
rs989918059 Chr.6: 33206530-33206530 on Build GRCh38
rs769214521 Chr.6: 33206441-33206441 on Build GRCh38
rs766047251 Chr.6: 33206354-33206354 on Build GRCh38
rs781491953 Chr.6: 33206414-33206414 on Build GRCh38
rs370933509 Chr.6: 33206348-33206348 on Build GRCh38

View Full Product Details